Skip to Content
Merck
All Photos(1)

Key Documents

SRP3065

Sigma-Aldrich

IGF-Binding Protein 6 human

recombinant, expressed in (BTI-Tn-5B1-4) High-5 Insect Cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

IBP-6

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$444.00

$444.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
20 μG
$444.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

$444.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in (BTI-Tn-5B1-4) High-5 Insect Cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

0.1-0.4 μg/mL

mol wt

23.0-30.0

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IGFBP6(3489)

Related Categories

General description

The IGFBP6 (insulin-like growth factor-binding protein 6) gene is mapped to human chromosome 12q13.[1] It belongs to the IGFBP family of proteins. The protein has a IGF (insulin-like growth factor)-binding site and a thyroglobulin type-1 domain.[2] IGFBPs are mainly required to regulate IGFs activities. IGFBP6 strongly interacts with IGF-2, but also interacts with IGF-1. It is also a hypoxia-inducible gene.[3][4]
Recombinant human IGF-BP6 has a calculated mass of 22.3kDa and consists of 213 amino acid residues including the IGF-BP domain and thyroglobulin type-I domain.

Biochem/physiol Actions

IGFBP6 (insulin-like growth factor-binding protein 6) can suppress growth of various IGF-2 (insulin-like growth factor 2)-dependent tumors. However, it also exhibits IGF-independent activities.[3] IGFBP is suggested as a serum biomarker for proliferative vitreoretinopathy.[5] HIV (human immunodeficiency virus) infection results in downregulation of IGFBP6 via differential methylation.[6]

Sequence

RCPGCGQGVQ AGCPGGCVEE EDGGSPAEGC AEAEGCLRRE GQECGVYTPN CAPGLQCHPP KDDEAPLRAL LLGRGRCLPA RAPAVAEENP KESKPQAGTA RPQDVNRRDQ QRNPGTSTTP SQPNSAGVQD TEMGPCRRHL DSVLQQLQTE VYRGAQTLYV PNCDHRGFYR KRQCRSSQGQ RRGPCWCVDR MGKSLPGSPD GNGSSSCPTG SSG

Physical form

Lyophilized from 10 mM Sodium Citrate, pH 3.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IGF binding protein-6 expression in vascular endothelial cells is induced by hypoxia and plays a negative role in tumor angiogenesis.
Zhang C, et al.
International Journal of Cancer. Journal International Du Cancer, 130, 2003-2003 (2012)
Leon A Bach
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society, 15(3), 185-192 (2005-05-26)
Insulin-like growth factor binding protein (IGFBP)-6 is unique among IGFBPs for its IGF-II binding specificity. IGFBP-6 inhibits growth of a number of IGF-II-dependent cancers, including rhabdomyosarcoma, neuroblastoma and colon cancer. Although the major action of IGFBP-6 appears to be inhibition
Whole genome methylation array reveals the down-regulation of IGFBP6 and SATB2 by HIV-1.
Zhang Y, et al.
Scientific Reports, 5, 10806-10806 (2015)
Kininogen 1 and insulin-like growth factor binding protein 6: candidate serum biomarkers of proliferative vitreoretinopathy.
Yu J, et al.
Clinical & Experimental Optometry : Journal of the Australian Optometrical Association, 97, 72-72 (2014)
Prohibitin-2 binding modulates insulin-like growth factor-binding protein-6 (IGFBP-6)-induced rhabdomyosarcoma cell migration.
Fu P, et al.
The Journal of Biological Chemistry, 288, 29890-29890 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service