Skip to Content
Merck
All Photos(2)

Key Documents

SRP2124

Sigma-Aldrich

Dr1 (NC2β, 19 kDa), GST tagged human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

NC2, NC2-beta

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$505.00

$505.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.


Select a Size

Change View
10 μG
$505.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$505.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~46.2 kDa

packaging

pkg of 10 μg

concentration

500 μg/mL

color

colorless to clear

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... DR1(1810)

Biochem/physiol Actions

Dr1 is a general negative regulator of class II and class III gene expression. It binds to the basic repeat domain of TBP on promoter DNA and can prevent the RNA polymerase II holoenzyme, or its TFIIB and/or TFIIA subunits, from assembling into an initiation complex. Dr1 is phosphorylated in vivo and this modification affects its interaction with TBP. In addition, Dr1 interacts with the hyperphosphorylated form of Pol II and with the repression domain of the AREB6 repressor. Dr1 forms a heterodimer complex with DRAP1 through its histone fold domain. More recently it was discovered that Dr1-DRAP1 is a bi-functional basal transcription factor that differentially regulates gene transcription through DPE (downstream promoter elements) or TATA box motifs. It can stimulate transcription in vitro from Drosophila promoters containing DPEs, whereas it represses transcription from TATA-containing promoters.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J White et al.
Science (New York, N.Y.), 266(5184), 448-450 (1994-10-21)
RNA polymerases I, II, and III each use the TATA-binding protein (TBP). Regulators that target this shared factor may therefore provide a means to coordinate the activities of the three nuclear RNA polymerases. The repressor Dr1 binds to TBP and
F Mermelstein et al.
Genes & development, 10(8), 1033-1048 (1996-04-15)
A Dr1-associated polypeptide (DRAP1) was isolated from HeLa cells and found to function as a corepressor of transcription. Corepressor function requires an interaction between DRAP1 and Dr1. Heterodimer formation was dependent on a histone fold motif present at the amino
J A Inostroza et al.
Cell, 70(3), 477-489 (1992-08-07)
We have discovered a protein termed Dr1 that interacts with the TATA-binding protein, TBP. The association of Dr1 with TBP results in repression of both basal and activated levels of transcription. The interaction of Dr1 with TBP precludes the formation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service