Skip to Content
Merck
All Photos(1)

Key Documents

SRP2058

Sigma-Aldrich

FXR-ligand binding domain (222-472) human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

FXR, Farnesol receptor HRR-1, NR1H4, retinoid receptor

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$410.00

$410.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
$410.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

$410.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~31.8 kDa

packaging

pkg of 10 μg

concentration

650 μg/mL

color

colorless to clear

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HRR-1(9971)

Biochem/physiol Actions

Farnesoid-X-activated receptor (FXR) was originally identified and cloned in rat as an orphan nuclear hormone receptor based on hybridization with a degenerate oligonucleotide designed from the highly conserved nuclear hormone receptor DNA binding domain. FXR functions as a heterodimer with RXR and binds to sequence elements in the promoters of target genes. The FXR/RXR heterodimer binds with highest affinity to inverted repeats separated by 1 bp (IR-1) and with low affinity to direct repeats separated by 4 and 5 bp (DR-4 and DR-5). As is the case for other nuclear hormone receptors, FXR regulates target gene activity in response to ligand. While initial studies suggested that farnesol and retinoid metabolites were likely ligands for FXR, current data support the notion that FXR is a bile acid sensor that plays an integral role in bile acid synthesis and transport. In the small intestine, FXR regulates bile acid uptake through the upregulation of the ileal bile acid binding protein gene via binding to an upstream response element. The FXR/RXR heterodimer can be activated by the bile salt chenodeoxycholic acid (CDCA) and FXR is required for the bile salt-dependent transcriptional control of the human ABCB11 gene (the bile salt export pump). In addition, FXR has been shown to inhibit the cholesterol 7-hydrolase gene (CYP7A1) transcription.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Seol et al.
Molecular endocrinology (Baltimore, Md.), 9(1), 72-85 (1995-01-01)
We have used a yeast genetic system to isolate cDNAs encoding proteins that specifically interact with the ligand-binding domain of human retinoid X receptor-alpha (RXR alpha). A number encoded portions of two known RXR heterodimer partners, the retinoic acid receptor
A M Zavacki et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(15), 7909-7914 (1997-07-22)
Retinoids are crucial regulators of a wide variety of processes in both developing and adult animals. These effects are thought to be mediated by the retinoic acid (RA) receptors and the retinoid X receptors (RXRs). We have identified an additional
B M Forman et al.
Cell, 81(5), 687-693 (1995-06-02)
Nuclear hormone receptors comprise a superfamily of ligand-modulated transcription factors that mediate the transcriptional activities of steroids, retinoids, and thyroid hormones. A growing number of related proteins have been identified that possess the structural features of hormone receptors, but that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service