Key Documents
TEM-CF150CU50
Sigma-Aldrich
Carbon Film Supported Copper Grid
Standard Thickness, size 150 mesh, box of 50
Synonym(s):
Carbon Film 150 Mesh, Copper
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-CF150CU50 is currently not available for purchase. Contact Technical Service
material
carbon film
copper grid
packaging
box of 50
film size
5-6 nm
size
150 mesh
General description
TEM grids supported by carbon film only. A 5-6 nm thin film of pure carbon deposited on one side of the standard square mesh grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)