Skip to Content
Merck
All Photos(1)

Key Documents

SRP0694

Sigma-Aldrich

TAU-441 (244-372) Protein

recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$555.00

$555.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
$555.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$555.00


Usually ships in 2 weeks.

recombinant

expressed in E. coli

Assay

70-95% (SDS-PAGE)

form

frozen liquid

mol wt

~15 kDa

concentration

0.2 μg/μL

accession no.

P10636-8

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAPT(4137)

Application

Useful for kinase assay and Western blot.

Biochem/physiol Actions

Recombinant human tag-free Tau-441 (244-372) was expressed in E. coli cells. Tau-441 or Tau-F is a member of the Tau family of proteins which function to stabilize the microtubules by binding to them. Tau proteins are subject to phosphorylation and this phenomenon regulates the association of the Tau protein with the microtubules (1). Deposits of Alzheimer′s disease AD-associated proteins, such as hyperphosphorylated Tau, as well as other shared misfolded proteins, such as, β-amyloid precursor protein (βAPP), ubiquitin, and various chaperones and protein kinases are thought to play a pathologic role in the cognitive decline and muscular failure. Malfunctioning of Tau proteins is associated with microtubules disintegration and collapsing of the neuronal transport system (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service