Skip to Content
Merck
All Photos(2)

Key Documents

SAE0076

Sigma-Aldrich

Tau-441 human

recombinant, lyophilized powder, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

Neurofibrillary tangle protein, PHF-tau, Paired helical filament-tau

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$708.00

$708.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$708.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$708.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 50.3 kDa (including c-terminal His and V5 tags)
observed mol wt 70-100 kDa by SDS-PAGE (The protein migrates as a 70-110 kDa protein on SDS-PAGE due to glycosylation)

technique(s)

cell based assay: suitable

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... MAPT(4137)

General description

Research area: Neuroscience

Tau-441 is a member of the Tau family of proteins. Tau is a family of neuronal microtubule-associated proteins. Six isoforms have been found that differ from each other in having either 3 or 4 binding repeats (R) of 31-32 amino acids, and from zero to 3 amino terminal inserts (N) of 29 amino acids each.

Application

Tau-441 human has been used to study the tau uptake by human bronchial epithelial cells (HBEC).[1]

Biochem/physiol Actions

Tau proteins are mainly expressed in the neurons of the central nervous system where they exert a role in stabilizing microtubules, key components of axonal transport, as well as in signal transduction. Tau proteins are subject to phosphorylation, which regulates their association with the microtubules. Deposits of Alzheimer′s disease AD-associated proteins, such as hyperphosphorylated Tau, as well as other shared misfolded proteins, such as beta-amyloid precursor protein (beta-APP), ubiquitin, and various chaperones and protein kinases, are thought to play a pathologic role in the cognitive decline and muscular failure. Malfunctioning of Tau proteins is associated with microtubule disintegration and collapsing of the neuronal transport system. Among other diseases, Tau forms in cerebrospinal fluid are considered a reliable biomarker for progressive supranuclear palsy, where the levels of Tau forms ratio were significantly reduced.

Sequence

MAEPRQEFEVMEDHAGTYGLGDRKDQGGYTMHQDQEGDTDAGLKESPLQTPTEDGSEEPGSETSDAKSTPTAEDVTAPLVDEGAPGKQAAAQPHTEIPEGTTAEEAGIGDTPSLEDEAAGHVTQARMVSKSKDGTGSDDKKAKGADGKTKIATPRGAAPPGQKGQANATRIPAKTPPAPKTPPSSGEPPKSGDRSGYSSPGSPGTPGSRSRTPSLPTPPTREPKKVAVVRTPPKSPSSAKSRLQTAPVPMPDLKNVKSKIGSTENLKHQPGGGKVQIINKKLDLSNVQSKCGSKDNIKHVPGGGSVQIVYKPVDLSKVTSKCGSLGNIHHKPGGGQVEVKSEKLDFKDRVQSKIGSLDNITHVPGGGNKKIETHKLTFRENAKAKTDHGAEIVYKSPVVSGDTSPRHLSNVSSTGSIDMVDSPQLATLADEVSASLAKQGLDRIRGRKLGPFEGKPIPNPLLGLDSTRTGHHHHHHHHGGQ

This sequence includes a polyhistidine and V5 tags at the C-terminus.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhang Zhenxia et al.
International journal of biological macromolecules, 171, 74-81 (2020-12-11)
In this study, the in vitro assembly of tau and anti-amyloidogenic properties of one naturally occurring phytoestrogen, calycosin, was investigated by spectroscopic techniques including ThT and ANS fluorescence, CD, Congo red absorbance as well as TEM analysis. Afterwards the cytotoxicity
Soluble pathogenic tau enters brain vascular endothelial cells and drives cellular senescence and brain microvascular dysfunction in a mouse model of tauopathy.
Hussong, et al.
Nature Communications, 14, 2367-2367 (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service