Skip to Content
Merck
All Photos(1)

Key Documents

SRP0193

Sigma-Aldrich

PARP2 Active human

recombinant, expressed in baculovirus infected insect cells, ≥60% (SDS-PAGE)

Synonym(s):

ADPRT2, ADPRTL3, NAD(+) ADP-ribosyltransferase 2, Poly (ADP-ribose) Polymerase 2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$922.00

$922.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
$922.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$922.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥60% (SDS-PAGE)

form

aqueous solution

mol wt

92 kDa

packaging

pkg of 10 μg

manufacturer/tradename

Sigma-Aldrich

concentration

>0.02 mg/mL

technique(s)

inhibition assay: suitable

solubility

water: soluble

NCBI accession no.

UniProt accession no.

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PARP2(10038)

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit of PARP incorporates 100 pmoles of poly(ADP) in 1 minute (room temperature) from NAD into acid-insoluble form.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PARP-2 domain requirements for DNA damage-dependent activation and localization to sites of DNA damage.
Riccio AA
Nucleic Acids Research, 44(4), 1691-1702 (2016)
PARP-2, A novel mammalian DNA damage-dependent poly(ADP-ribose) polymerase.
Ame JC
The Journal of Biological Chemistry, 274(25), 17860-17868 (1999)
PARP-2 regulates cell cycle-related genes through histone deacetylation and methylation independently of poly(ADP-ribosyl)ation.
Liang YC
Biochemical and Biophysical Research Communications, 431(1), 58-64 (2013)
Xiao-Nan Zhang et al.
Nature communications, 10(1), 4196-4196 (2019-09-15)
Nicotinamide adenine dinucleotide (NAD+)-dependent ADP-ribosylation plays important roles in physiology and pathophysiology. It has been challenging to study this key type of enzymatic post-translational modification in particular for protein poly-ADP-ribosylation (PARylation). Here we explore chemical and chemoenzymatic synthesis of NAD+
Sharon McGonigle et al.
Oncotarget, 6(38), 41307-41323 (2015-10-30)
Inhibition of Poly(ADP-ribose) Polymerase1 (PARP1) impairs DNA damage repair, and early generation PARP1/2 inhibitors (olaparib, niraparib, etc.) have demonstrated clinical proof of concept for cancer treatment. Here, we describe the development of the novel PARP inhibitor E7449, a potent PARP1/2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service