Skip to Content
Merck
All Photos(1)

Key Documents

P0996

Sigma-Aldrich

PARP-1 human

recombinant, expressed in E. coli

Synonym(s):

NAD+ ADP-ribosyltransferase, Poly(ADP-ribose) Polymerase

Sign Into View Organizational & Contract Pricing

Select a Size

1000 UNITS
$817.00

$817.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1000 UNITS
$817.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

$817.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli

form

solution

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PARP1(142)

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ADP-ribose) polymerase 1 (PARP-1) is associated with the inflammation response during atherosclerosis. [1]
Poly(ADP-ribose) polymerase 1 (PARP1) belongs to the PARP family which is highly conserved during evolution. PARP1 gene is localized at 1q42.12 in the human chromosome.[2]

Application

PARP-1 human has been used in the GST-pull down assay for immunoprecipitation of Glutathione S-transferase (GST) or GST- cyclic GMP- AMP synthase (cGAS) (GST-cGAS).[3] It has also been used in in vitro ribosylation assay.[4]
PARP-1 is a nuclear enzyme that synthesizes ADP-ribose polymers from NAD+, specifically binds Zn2+ and DNA, and recognizes single-strand breaks in DNA. PARP1 has been used in a study to assess racial and tissue-specific cancer risk associated with polymorphism in the PARP1 gene. [5] It has also been used in a study to investigate inhibitors of PARP-1 for potential cancer treatments. [6]

Biochem/physiol Actions

PARP-1 is inactivated by cleavage into a 24kDA and 89kDA fragment by activated caspase-3 or caspase-7. This results in the decreased ability to repair DNA damage and an increase in apoptosis. [7]
PARP-1, a nuclear enzyme that synthesizes ADP-ribose polymers from NAD, specifically binds Zn2+ and DNA, and recognizes single-strand breaks in DNA. It is involved in base excision repair, both short-patch and long-patch, rejoining DNA strand breaks and plays a role in p53 expression and activation. A high level of basal neuronal DNA damage and PARP activity has been reported in rat brain tissue. PARP-1 was shown to be required for HIV-1 integration into DNA. If PARP-1 is deficient there is no productive HIV-1 infection.
Poly(ADP-ribose) polymerase 1 (PARP1) is essential for diverse functions like DNA damage detection and repair, chromatin modification, cell differentiation, transcription and apoptotic cell death. It also plays a major role in spermatogenesis. Polymorphism in the gene PARP1 is associated with oligospermia and causes male infertility.[2]

Unit Definition

One unit will incorporate 10 femptomole of poly(ADP-ribose) from NAD into 5 μg of immobilized histone proteins in 30 minutes at 22 °C in a 96 well plate.

Physical form

Supplied as a solution in 20 mM Tris-HCl, pH 8.0, 200 mM NaCl, 1 mM DTT, 0.1% Triton-X 100, 50 % glycerol, and 0.1 mg/ml BSA.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear cGAS suppresses DNA repair and promotes tumorigenesis
Liu H, et al.
Nature, 563(7729), 131-131 (2018)
S Muller et al.
Clinical immunology and immunopathology, 73(2), 187-196 (1994-11-01)
Poly(ADP-Ribose) polymerase (PARP) is a chromatin-associated enzyme that specifically binds to DNA strand breaks in a zinc-dependent manner. We describe here the presence of IgG antibodies reacting with recombinant human PARP in the serum of patients with systemic lupus erythematosus
David Davidson et al.
Investigational new drugs, 31(2), 461-468 (2012-10-12)
Poly [ADP-ribose] polymerase-1 (PARP-1) localizes rapidly to sites of DNA damage and has been associated with various repair mechanisms including base excision repair (BER) and homologous recombination/non-homologous end joining (HRR/NHEJ). PARP-1 acts by adding poly-ADP ribose side chains to target
PARP cleavage as a means of assessing apoptosis.
Peter Mullen
Methods in molecular medicine, 88, 171-181 (2003-11-25)
E Babiychuk et al.
The Plant journal : for cell and molecular biology, 15(5), 635-645 (1998-10-21)
One of the immediate reactions of the mammalian cell to many environmental stresses is a massive synthesis of poly(ADP-ribose), catalyzed by poly(ADP-ribose) polymerase (PARP). Most of the biological functions attributed to PARP are inferred from experimentation with mammalian cells. In

Questions

  1. What is the total weight of the recombinant protein for 1000 units?

    1 answer
    1. The total weight can be determined using the Unit Activity and the Protein Concentration reported on the lot specific Certificate of Analysis. For example, for the 1KU pack size, if the activity is 10 units/µl and the protein concentration is 0.384 mg/ml the weight would be 0.26mg.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service