SRP0168
EED human
recombinant, expressed in baculovirus infected insect cells, ≥90% (SDS-PAGE)
Synonym(s):
HEED, Rinchik 5, WAIT1, WD protein associating with integrin cytoplasmic tails 1, embryonic ectoderm development
About This Item
Recommended Products
biological source
human
recombinant
expressed in baculovirus infected insect cells
Assay
≥90% (SDS-PAGE)
form
aqueous solution
mol wt
51 kDa
packaging
pkg of 50 μg
concentration
>0.02 mg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... EED(8726)
General description
Human Embryonic Ectoderm Development (hEED), also known as WAIT-1, GenBank Accession No. NM_003797, amino acids 2-end with N-terminal FLAG-tag, MW = 51kDa, expressed in Sf9 cells via a baculovirus expression system.
Biochem/physiol Actions
Physical form
Preparation Note
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service