Skip to Content
Merck
All Photos(1)

Key Documents

SRP0159

Sigma-Aldrich

Histone H3 peptide (1-21) human

≥90% (HPLC)

Synonym(s):

HIST1H3E, Histone H3.1

Sign Into View Organizational & Contract Pricing

Select a Size

160 NMOL
$372.00

$372.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
160 NMOL
$372.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$372.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

Assay

≥90% (HPLC)

form

aqueous solution

mol wt

~2 kDa

packaging

pkg of 160 nmol

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HIST1H3E(8353)

Application

Study enzyme kinetics, and screen small molecular inhibitors of Histone 3, K4,K9, methyltransferases for drug discovery and HTS applications

Physical form

Supplied as water solution, 800 μM

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube to recover full content of the tube. Aliquot into single use aliquots. Store remaining product in aliquots at -70°C. Note: Avoid freeze/thaw cycles.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

PTMs on H3 variants before chromatin assembly potentiate their final epigenetic state.
Loyola A
Molecular Cell, 24(2), 309-316 (2006)
The differential mobilization of histones H3.1 and H3.3 by herpes simplex virus 1 relates histone dynamics to the assembly of viral chromatin.
Conn KL
PLoS Pathogens, 9(10), e1003695-e1003695 (2013)
The program for processing newly synthesized histones H3.1 and H4.
Campos EI
Nature Structural and Molecular Biology, 17(11), 1343-1351 (2010)
Hideaki Tagami et al.
Cell, 116(1), 51-61 (2004-01-14)
Deposition of the major histone H3 (H3.1) is coupled to DNA synthesis during DNA replication and possibly DNA repair, whereas histone variant H3.3 serves as the replacement variant for the DNA-synthesis-independent deposition pathway. To address how histones H3.1 and H3.3
The histone variant H3.3 marks active chromatin by replication-independent nucleosome assembly.
Ahmad K and Henikoff S
Molecular Cell, 9(6), 1191-1200 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service