Skip to Content
Merck
All Photos(1)

Documents

H4380

Sigma-Aldrich

Histone from calf thymus

Type VIII-S

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1895465
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51

biological source

bovine (calf) thymus

type

Type VIII-S

form

powder

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Do not confuse Sigma "Type" designations with the "Fraction" designations used by Luck and co-workers.

Quality

Contains other subgroups

Preparation Note

Arginine-rich subgroup f3 isolated by a modification of the method of Johns, E.W., Biochem. J., 92, 55 (1964).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renata de Moraes Maciel et al.
Biochemical and biophysical research communications, 370(1), 53-56 (2008-03-19)
The transcriptional co-activator GCN5, a histone acetyltransferase (HAT), is part of large multimeric complexes that are required for chromatin remodeling and transcription activation. As in other eukaryotes, the DNA from the parasite Schistosome mansoni is organized into nucleosomes and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service