Skip to Content
Merck
All Photos(1)

Key Documents

SML3435

Sigma-Aldrich

Momordin Ιc

≥98% (HPLC)

Synonym(s):

(3β)-17-Carboxy-28-norolean-12-en-3-yl 3-O-β-D-xylopyranosyl-β-D-glucopyranosiduronic acid, Mormordin Ic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H64O13
CAS Number:
Molecular Weight:
764.94
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 12 to 17° in methanol (c=0.5 g/100mL)

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1C(=O)O)O)O[C@@H]7OC[C@H]([C@@H]([C@H]7O)O)O)O)O[C@@H]2C([C@H]3[C@@]([C@@H]4[C@]([C@@]5(CC[C@@]6([C@@H](CC(CC6)(C)C)C5=CC4)C(=O)O)C)(CC3)C)(CC2)C)(C)C

InChI

1S/C41H64O13/c1-36(2)14-16-41(35(49)50)17-15-39(6)20(21(41)18-36)8-9-24-38(5)12-11-25(37(3,4)23(38)10-13-40(24,39)7)52-34-29(46)30(28(45)31(54-34)32(47)48)53-33-27(44)26(43)22(42)19-51-33/h8,21-31,33-34,42-46H,9-19H2,1-7H3,(H,47,48)(H,49,50)/t21-,22+,23-,24+,25-,26-,27+,28-,29+,30-,31-,33-,34+,38-,39+,40+,41-/m0/s1

InChI key

HWYBGIDROCYPOE-WEAQAMGWSA-N

General description

Momordin Ιc is a pentacyclic triterpenoid isolated from Kochia scoparia (L.) fruit and other medicinal plants.

Biochem/physiol Actions

Momordin Ic is a potent inhibitor of SUMO-specific protease 1 (SENP1) that increases SUMOylated proteins in PC3 cells. It inhibits cell proliferation and induces cell death in prostate cancer PC3 xenograft mouse model. Momordin Ιc decreases serum glucose and ethanol levels after oral administration through acceleration of gastrointestinal transit.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Small-Molecule Inhibitors Targeting Protein SUMOylation as Novel Anticancer Compounds
Yang Y, Xia Z, Wang X, Zhao X, Sheng Z, Ye Y, He G, Zhou L, Zhu H, Xu N, Liang S
Molecular Pharmacology (2018)
Y Li et al.
European journal of pharmacology, 392(1-2), 71-77 (2000-04-05)
Possible involvement of 5-hydroxytryptamine (5-HT), 5-HT receptors and prostaglandins in the acceleration of gastrointestinal transit by momordin Ic was investigated in mice. Accelerative effect of momordin Ic (25 mg/kg, p.o.) on gastrointestinal transit was attenuated by pretreatment with a bolus
Jingjing Wu et al.
Oncotarget, 7(37), 58995-59005 (2016-07-28)
SUMO-specific protease 1 (SENP1), a member of the de-SUMOylation protease family, is elevated in prostate cancer (PCa) cells and is involved in PCa pathogenesis. Momordin Ιc (Mc), a natural pentacyclic triterpenoid, inhibited SENP1 in vitro, as reflected by reduced SENP1C-induced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service