The GST tag is located at the N terminus of the protein. unable to share whether this protein contains a linker or not as this is considered proprietary.
S0439
SUMO-1 human
≥95% (SDS-PAGE), recombinant, expressed in E. coli (GST-tagged)
Synonym(s):
Sentrin-1
Select a Size
$529.00
Select a Size
About This Item
$529.00
Recommended Products
recombinant
expressed in E. coli (GST-tagged)
Assay
≥95% (SDS-PAGE)
mol wt
38.5 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... SUMO1(7341)
General description
Application
Biochem/physiol Actions
Physical form
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is S0439-1MG (SUMO-1 human) N- or C-terminal GST tagged? Is there any form of linker amino acids between the SUMO-1 sequence and the GST sequence?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service