Skip to Content
Merck
All Photos(1)

Key Documents

SML3148

Sigma-Aldrich

BI8622

≥98% (HPLC)

Synonym(s):

BI 8622, BI-8622, N-[4-(Aminomethyl)phenyl]-6-(4-cyano-4-phenyl-1-piperidinyl)-5-methyl-4-pyrimidinecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$151.00
25 MG
$605.00

$151.00


Estimated to ship onMay 05, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$151.00
25 MG
$605.00

About This Item

Empirical Formula (Hill Notation):
C25H26N6O
CAS Number:
Molecular Weight:
426.51
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.77

$151.00


Estimated to ship onMay 05, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N#CC1(CCN(CC1)C2=C(C)C(C(NC3=CC=C(CN)C=C3)=O)=NC=N2)C4=CC=CC=C4

InChI

1S/C25H26N6O/c1-18-22(24(32)30-21-9-7-19(15-26)8-10-21)28-17-29-23(18)31-13-11-25(16-27,12-14-31)20-5-3-2-4-6-20/h2-10,17H,11-15,26H2,1H3,(H,30,32)

InChI key

IJHAXMJRQQTBPL-UHFFFAOYSA-N

Biochem/physiol Actions

BI8622 is a selective HUWE1 (HECTH9; MULE; ARF-BP1) inhibitor (IC50 = 3.2 μM by HUWE1 HECT-domain auto-ubiquitination assay; IC50 >50 μM against other HECT-domain Ub ligases tested). BI8622 inhibits the degradation of HUWE1 target proteiins, including MCL1 (IC50 = 6.8 μM; UV irradiated HeLa MCL1 transfectants), MIZ1 (10 μM; Ls174T), and TopBP1. BI8622 suppresses Ls174T colony formation (IC50 = 8.4 μM in 5 days) and the growth of three colon carcinoma cell lines (20 μM for 5 days; HCT116, HT29, SW480).
Selective HUWE1 (HECTH9; MULE; ARF-BP1) inhibitor against HUWE1-mediated degration of target proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aswini Krishnan et al.
Nature communications, 11(1), 2056-2056 (2020-04-30)
Papillary thyroid cancer (PTC) is the most common type of endocrine malignancy. By RNA-seq analysis, we identify a RET rearrangement in the tumour material of a patient who does not harbour any known RAS or BRAF mutations. This new gene
Yu Guo et al.
The Journal of clinical investigation, 130(12), 6301-6316 (2020-10-27)
The mechanism by which inflammasome activation is modulated remains unclear. In this study, we identified an AIM2-interacting protein, the E3 ubiquitin ligase HUWE1, which was also found to interact with NLRP3 and NLRC4 through the HIN domain of AIM2 and
Hye In Cho et al.
Cell death and differentiation, 27(12), 3273-3288 (2020-06-20)
Brpf-histone acetyltransferase (HAT) complexes have important roles in embryonic development and regulating differentiation in ESCs. Among Brpf family, Brpf3 is a scaffold protein of Myst2 histone acetyltransferase complex that plays crucial roles in gene regulation, DNA replication, development as well
Viktoria Kunz et al.
Scientific reports, 10(1), 18419-18419 (2020-10-30)
Experimental evidence suggests that ubiquitin-protein ligases regulate a number of cellular processes involved in tumorigenesis. We analysed the role of the E3 ubiquitin-protein ligase HUWE1 for pathobiology of multiple myeloma (MM), a still incurable blood cancer. mRNA expression analysis indicates
Stefanie Peter et al.
EMBO molecular medicine, 6(12), 1525-1541 (2014-09-26)
Deregulated expression of MYC is a driver of colorectal carcinogenesis, necessitating novel strategies to inhibit MYC function. The ubiquitin ligase HUWE1 (HECTH9, ARF-BP1, MULE) associates with both MYC and the MYC-associated protein MIZ1. We show here that HUWE1 is required

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service