Skip to Content
Merck
All Photos(1)

Documents

SML2920

Sigma-Aldrich

Y-39983 dihydrochloride

≥98% (HPLC)

Synonym(s):

4-[(1R)-1-Aminoethyl]-N-1H-pyrrolo[2,3-b]pyridin-4-yl-benzamide dihydrochloride, Y 39983, Y39983

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N4O · 2HCl
CAS Number:
Molecular Weight:
353.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +5 to +9°, c = 1 in methanol

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

N[C@@H](C1=CC=C(C(NC2=CC=NC3=C2C=CN3)=O)C=C1)C.Cl.Cl

InChI

1S/C16H16N4O.2ClH/c1-10(17)11-2-4-12(5-3-11)16(21)20-14-7-9-19-15-13(14)6-8-18-15;;/h2-10H,17H2,1H3,(H2,18,19,20,21);2*1H/t10-;;/m1../s1

InChI key

CKFHAVRPVZNMGT-YQFADDPSSA-N

Biochem/physiol Actions

Y-39983 is a potent and selective Rho-kinase (ROCK) inhibitor (IC50s = 3.6 nM). Y-39983 is 10 fold more potent in promoting the outgrowth of neurites in cultured retinal slices and axonal regeneration in crushed optic nerve models than Y-27632. Y-39983 act also as an activator of the neuroprotective Parkin-mediated mitophagy pathway.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin G Chen et al.
Journal of visualized experiments : JoVE, (89) (2014-08-01)
Human pluripotent stem cells (hPSCs) hold great promise for regenerative medicine and biopharmaceutical applications. Currently, optimal culture and efficient expansion of large amounts of clinical-grade hPSCs are critical issues in hPSC-based therapies. Conventionally, hPSCs are propagated as colonies on both
Enhua H Zhou et al.
Investigative ophthalmology & visual science, 58(7), 2991-3003 (2017-06-13)
To discover novel therapies that lower IOP by increasing aqueous humor outflow facility, ex vivo ocular perfusion systems provide a valuable tool. However, currently available designs are limited by their throughput. Here we report the development of a compact, scalable
Natalia Moskal et al.
Nature communications, 11(1), 88-88 (2020-01-05)
The accumulation of damaged mitochondria causes the death of dopaminergic neurons. The Parkin-mediated mitophagy pathway functions to remove these mitochondria from cells. Targeting this pathway represents a therapeutic strategy for several neurodegenerative diseases, most notably Parkinson's disease. We describe a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service