Skip to Content
Merck
All Photos(1)

Key Documents

SML3131

Sigma-Aldrich

GSK620

≥98% (HPLC)

Synonym(s):

1-Benzyl-N5-cyclopropyl-N3-methyl-2-oxo-1,2-dihydropyridine-3,5-dicarboxamide, GSK 620, GSK-620, N5-Cyclopropyl-1,2-dihydro-N3-methyl-2-oxo-1-(phenylmethyl)-3,5-pyridinedicarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$240.00
25 MG
$965.00

$240.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$240.00
25 MG
$965.00

About This Item

Empirical Formula (Hill Notation):
C18H19N3O3
CAS Number:
Molecular Weight:
325.36
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$240.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N(C=C(C=C1C(NC)=O)C(NC2CC2)=O)CC3=CC=CC=C3

InChI key

QZZCUOVXHPAQRQ-UHFFFAOYSA-N

Biochem/physiol Actions

GSK620 Is an orally active, potent and selective pan-second bromodomain (BD2) inhibitor (BD2/BD1 pIC50 = 6.5/4.6/BRD2, 7.1/4.6/BRD3, 7.1/4.8/BRD4, 6.7/4.6/BRDT) with improved oral availability than GSK046 (iBET-BD2) and in vivo efficacy in inflammatory disease models, including collagen-induced arthritis (3-10 mg/kg/d p.o. in rats), imiquimod (IMQ)-induced psoriasis (20 mg/kg/d p.o. in mice), and a nonalcoholic steatohepatitis (NAFLD) model (20 mg/kg/d p.o. in STAM mice).
Potent and selective pan-second bromodomain (BD2) inhibitor with in vivo efficacy in inflammatory disease models and improved oral availability than GSK046.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service