Skip to Content
Merck
All Photos(1)

Key Documents

SML2192

Sigma-Aldrich

A-485

≥98% (HPLC)

Synonym(s):

N-(4-fluorobenzyl)-2-((R)-5-(3-methylureido)-2′,4′-dioxo-2,3-dihydrospiro[indene-1,5′-ozazolidine]-3′-7 yl)-N-((S)-1,1,1-trifluoropropan-2-yl)acetamide, Spiro[1H-indene-1,5′-oxazolidine]-3′-acetamide, N-[(4-fluorophenyl)methyl]-2,3-dihydro-5-[[(methylamino)carbonyl]amino]-2′,4′-dioxo-N-[(1S)-2,2,2-trifluoro-1-methylethyl]-, (1R)-

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$242.00
25 MG
$966.00

$242.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$242.00
25 MG
$966.00

About This Item

Empirical Formula (Hill Notation):
C25H24F4N4O5
CAS Number:
Molecular Weight:
536.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$242.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +40 to +50°, c = 0.5 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CNC(NC1=CC=C2C(CC[C@]23C(N(CC(N(CC4=CC=C(F)C=C4)[C@@H](C)C(F)(F)F)=O)C(O3)=O)=O)=C1)=O

InChI

1S/C25H24F4N4O5/c1-14(25(27,28)29)32(12-15-3-5-17(26)6-4-15)20(34)13-33-21(35)24(38-23(33)37)10-9-16-11-18(7-8-19(16)24)31-22(36)30-2/h3-8,11,14H,9-10,12-13H2,1-2H3,(H2,30,31,36)/t14-,24+/m0/s1

InChI key

VRVJKILQRBSEAG-LFPIHBKWSA-N

Application

A-485 has been used as a p300 regulated RelA acetylation inhibitor to study the effect of fatty acid oxidation (FAO) derived cytoplasmic acetyl-CoA on CD47 transcription and protein stability via citrate/acetyl-CoA mediated RelA acetylation in glioblastoma.[1]

Biochem/physiol Actions

A-485 exhibits an anti-tumor effect and may show therapeutic effects against the growth hormone pituitary adenoma (GHPA). It can specifically block the acetylation of histone H3 lysine 27 (H3K27) and lysine 18 (H3K18) sites catalyzed by p300.[2]
A-485 is a potent and selective catalytic inhibitor of the p300 and CREB-binding protein (CBP) histone acetyltransferases (HATs) with an IC50 value of 60 nM. A-485 was most active against haematological malignancies including most multiple myeloma cell lines as well as androgen receptor-positive prostate cancer. It was also shown to inhibit the proliferation of castration-resistant prostate cancer cells and in vivo tumour growth in SCID mice.
Selective catalytic p300/CBP inhibitor that targets lineage-specific tumours

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loren M Lasko et al.
Nature, 550(7674), 128-132 (2017-09-28)
The dynamic and reversible acetylation of proteins, catalysed by histone acetyltransferases (HATs) and histone deacetylases (HDACs), is a major epigenetic regulatory mechanism of gene transcription and is associated with multiple diseases. Histone deacetylase inhibitors are currently approved to treat certain
Chenxing Ji et al.
The Journal of clinical endocrinology and metabolism, 107(6), e2291-e2300 (2022-03-06)
Growth hormone pituitary adenoma (GHPA), a major subtype of pituitary adenoma (PA), can lead to progressive somatic disfigurement, multiple complications, and even increased mortality. The efficacy of current treatments is limited; thus, a novel pharmacological treatment is urgently needed. As
Nian Jiang et al.
Nature communications, 13(1), 1511-1511 (2022-03-23)
Glioblastoma multiforme (GBM) remains the top challenge to radiotherapy with only 25% one-year survival after diagnosis. Here, we reveal that co-enhancement of mitochondrial fatty acid oxidation (FAO) enzymes (CPT1A, CPT2 and ACAD9) and immune checkpoint CD47 is dominant in recurrent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service