Skip to Content
Merck
All Photos(1)

Key Documents

SML3125

Sigma-Aldrich

LEI-401

≥98% (HPLC)

Synonym(s):

N-(Cyclopropylmethyl)-6-((S)-3-hydroxypyrrolidin-1-yl)-2-((S)-3-phenylpiperidin-1-yl)pyrimidine-4-carboxamide, N-(Cyclopropylmethyl)-6-[(3S)-3-hydroxy-1-pyrrolidinyl]-2-[(3S)-3-phenyl-1-piperidinyl]-4-pyrimidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31N5O2
CAS Number:
Molecular Weight:
421.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC(N2CC[C@@H](C2)O)=NC(N3CCC[C@H](C3)C4=CC=CC=C4)=N1)NCC5CC5

InChI

1S/C24H31N5O2/c30-20-10-12-28(16-20)22-13-21(23(31)25-14-17-8-9-17)26-24(27-22)29-11-4-7-19(15-29)18-5-2-1-3-6-18/h1-3,5-6,13,17,19-20,30H,4,7-12,14-16H2,(H,25,31)/t19-,20+/m1/s1

InChI key

GFHJYPQZBBHOCC-UXHICEINSA-N

Biochem/physiol Actions

CNS-active, potent and selective inhibitor of N-acylphosphatidylethanolamine phospholipase D (NAPE-PLD)
LEI-401 is a CNS-active, potent and selective inhibitor of N-acylphosphatidylethanolamine phospholipase D (NAPE-PLD). LEI-401 reduces N-acylethanolamines (NAEs) in in the brain of freely moving mice and neuroblastoma cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elliot D Mock et al.
Journal of medicinal chemistry, 64(1), 481-515 (2021-01-01)
N-Acylphosphatidylethanolamine phospholipase D (NAPE-PLD) is regarded as the main enzyme responsible for the biosynthesis of N-acylethanolamines (NAEs), a family of bioactive lipid mediators. Previously, we reported N-(cyclopropylmethyl)-6-((S)-3-hydroxypyrrolidin-1-yl)-2-((S)-3-phenylpiperidin-1-yl)pyrimidine-4-carboxamide (1, LEI-401) as the first potent and selective NAPE-PLD inhibitor that decreased NAEs
Elliot D Mock et al.
Nature chemical biology, 16(6), 667-675 (2020-05-13)
N-acylethanolamines (NAEs), which include the endocannabinoid anandamide, represent an important family of signaling lipids in the brain. The lack of chemical probes that modulate NAE biosynthesis in living systems hamper the understanding of the biological role of these lipids. Using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service