Skip to Content
Merck
All Photos(1)

Key Documents

SML3072

Sigma-Aldrich

8-pCPT-2′-O-Me-cAMP-AM

≥98% (HPLC)

Synonym(s):

8-pCPT-2′-O-Me-cAMP-AM, 007-AM, 8-(4-Chlorophenylthio)-2′-O-methyladenosine-3′,5′-cyclic monophosphate, acetoxymethyl ester, 8-CPT-2′-O-Me-cAMP-AM, 8-CPT-AM, 8-pCPT-AM, CPT-AM

Sign Into View Organizational & Contract Pricing

Select a Size

1 μMOL
$510.00

$510.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 μMOL
$510.00

About This Item

Empirical Formula (Hill Notation):
C20H21ClN5O8PS
CAS Number:
Molecular Weight:
557.90
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$510.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

film or powder

color

colorless to light yellow

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=C(N2[C@H](O[C@H](CO3)[C@H]4OP3(OCOC(C)=O)=O)[C@@H]4OC)SC5=CC=C(C=C5)Cl

InChI

1S/C20H21ClN5O8PS/c1-10(27)30-9-32-35(28)31-7-13-15(34-35)16(29-2)19(33-13)26-18-14(17(22)23-8-24-18)25-20(26)36-12-5-3-11(21)4-6-12/h3-6,8,13,15-16,19H,7,9H2,1-2H3,(H2,22,23,24)/t13-,15-,16-,19-,35?/m1/s1

InChI key

FZMWUFYPEVDWPA-SILPBKOMSA-N

Biochem/physiol Actions

8-pCPT-2′-O-Me-cAMP-AM (CPT-AM) is a membrane-permeant acetoxymethyl (AM) ester precusor form of the EPAC-selective cAMP analog (ESCA) 8-pCPT-2′-O-Me-cAMP (CPT). CPT-AM is >100-times more cell-permeable than CPT (by FRET using CFP-Epac-YFP-expressing A431; No effect by PKA FRET assay) and >100-times more efficient in stimulating cellular Rap1 activation (ECmax = 0.1 μM/CPT-AM vs. 100 μM/CPT; HUVECs).
Acetoxymethyl (AM) ester precusor of the EPAC-selective cAMP analog 8-pCPT-2′-O-Me-cAMP with greatly improved membrane permeability and cellular efficacy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service