Skip to Content
Merck
All Photos(1)

Key Documents

B2557

Sigma-Aldrich

8-Bromo-2′-monobutyryladenosine-3′,5′-cyclic monophosphorothioate, Rp-isomer

>97% (HPLC), solid

Synonym(s):

Rp-8-Br-MB-cAMPS

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$1,200.00

$1,200.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Select a Size

Change View
1 VIAL
$1,200.00

About This Item

Empirical Formula (Hill Notation):
C14H16BrN5O6PSNa
Molecular Weight:
516.24
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

$1,200.00


Estimated to ship onMay 20, 2025


Request a Bulk Order

Quality Level

Assay

>97% (HPLC)

form

solid

packaging

vial of 5 μmol (2.5 mg solid)

solubility

water: soluble

storage temp.

−20°C

SMILES string

CCCC(=O)O[C@@H]1[C@@H]2O[P@](O)(=S)OC[C@H]2O[C@H]1n3c(Br)nc4c(N)ncnc34

InChI

1S/C14H17BrN5O6PS/c1-2-3-7(21)25-10-9-6(4-23-27(22,28)26-9)24-13(10)20-12-8(19-14(20)15)11(16)17-5-18-12/h5-6,9-10,13H,2-4H2,1H3,(H,22,28)(H2,16,17,18)/t6-,9-,10-,13-,27-/m1/s1

InChI key

KWTXVSKODFLFLA-WJCFKVOZSA-N

Application

Lipophilic precursor that is more lipophilic and membrane-permeable compared to Rp-8-Br-cAMPS.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Ruiz-Velasco et al.
Circulation research, 82(5), 557-565 (1998-04-07)
Cyclic nucleotides are known to modify voltage-gated (L-type) Ca2+ channel activity in vascular smooth muscle cells, but the exact mechanism(s) underlying these effects is not well defined. The purpose of the present study was to investigate the modulatory role of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service