Skip to Content
Merck
All Photos(1)

Key Documents

SML3070

Sigma-Aldrich

PSEM89S Hydrochloride salt

≥98% (HPLC)

Synonym(s):

(S)-2,5-Dimethoxy-N-(quinuclidin-3-yl)benzamide, Hydrochloride salt, N-(3S)-1-Azabicyclo[2.2.2]oct-3-yl-2,5-dimethoxybenzamide Hydrochloride salt, PSEM 89S Hydrochloride salt, PSEM-89S Hydrochloride salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$112.00
25 MG
$449.00

$112.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$112.00
25 MG
$449.00

About This Item

Empirical Formula (Hill Notation):
C16H22N2O3 · HCl
CAS Number:
Molecular Weight:
326.82
UNSPSC Code:
12352200
NACRES:
NA.77

$112.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N[C@@H]1CN2CCC1CC2)C3=C(C=CC(OC)=C3)OC.Cl

InChI

1S/C16H22N2O3.ClH/c1-20-12-3-4-15(21-2)13(9-12)16(19)17-14-10-18-7-5-11(14)6-8-18;/h3-4,9,11,14H,5-8,10H2,1-2H3,(H,17,19);1H/t14-;/m1./s1

InChI key

OHMFVSNTNXNVRE-PFEQFJNWSA-N

Biochem/physiol Actions

PSEM agonist for chimeric ligand-gated ion channel pores with nAChR α7 LBD L141F or L141F/Y115F mutant (PSAM) in vitro and in vivo.
PSEM89S is a pharmacologically selective effector molecule (PSEM) that acts as a selective agonist in vitro and in vivo for chimeric pharmacologically selective actuator modules (PSAMs) composed of nAChR α7 ligand-binding domain (LBD) with L141F mutation fused to the ion pore domain (IPD) of a ligand-gated ion channel (LGIC), including glycine receptor (PSAML141F-GlyR & PSAML141F/Y115F-GlyR). 5-HT3 (PSAML141F-5-HT3 & PSAML141F/Y115F-5-HT3) and high-conductance 5-HT3 (PSAML141F-5-HT3 HC & PSAML141F/Y115F-5-HT3 HC).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Nicholas Betley et al.
Nature, 521(7551), 180-185 (2015-04-29)
Homeostasis is a biological principle for regulation of essential physiological parameters within a set range. Behavioural responses due to deviation from homeostasis are critical for survival, but motivational processes engaged by physiological need states are incompletely understood. We examined motivational
Serena Cerritelli et al.
PloS one, 11(4), e0153187-e0153187 (2016-04-15)
Opioids are widely used medicinally as analgesics and abused for hedonic effects, actions that are each complicated by substantial risks such as cardiorespiratory depression. These drugs mimic peptides such as β-endorphin, which has a key role in endogenous analgesia. The
Anna L Vlasits et al.
Neuron, 83(5), 1172-1184 (2014-08-27)
Direction-selective ganglion cells (DSGCs) are tuned to motion in one direction. Starburst amacrine cells (SACs) are thought to mediate this direction selectivity through precise anatomical wiring to DSGCs. Nevertheless, we previously found that visual adaptation can reverse DSGCs's directional tuning
Wenjie Ren et al.
Nature neuroscience, 19(2), 220-222 (2015-12-23)
We examined adaptations in nucleus accumbens (NAc) neurons in mouse and rat peripheral nerve injury models of neuropathic pain. Injury selectively increased excitability of NAc shell indirect pathway spiny projection neurons (iSPNs) and altered their synaptic connectivity. Moreover, injury-induced tactile
Stefan Hirschberg et al.
eLife, 6 (2017-10-14)
The locus coeruleus (LC) projects throughout the brain and spinal cord and is the major source of central noradrenaline. It remains unclear whether the LC acts functionally as a single global effector or as discrete modules. Specifically, while spinal-projections from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service