Skip to Content
Merck
All Photos(1)

Key Documents

609021

Sigma-Aldrich

L-Lysine-15N2 hydrochloride

98 atom % 15N, 98% (CP)

Synonym(s):

(S)-2,6-Diaminocaproic acid-15N2 hydrochloride, (S)-2,6-Diaminohexanoic acid-15N2 hydrochloride, 15N Labeled lysine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$992.00

$992.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$992.00

About This Item

Linear Formula:
H215N(CH2)4CH(15NH2)CO2H · HCl
CAS Number:
Molecular Weight:
184.64
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

$992.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % 15N

Assay

98% (CP)

form

solid

optical activity

[α]20/D +21°, c = 8 in 5 M HCl

technique(s)

bio NMR: suitable
protein expression: suitable

mp

263-264 °C (dec.) (lit.)

mass shift

M+2

SMILES string

Cl[H].[15NH2]CCCC[C@H]([15NH2])C(O)=O

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1/i7+1,8+1;

InChI key

BVHLGVCQOALMSV-XVXIHWTMSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service