Skip to Content
Merck
All Photos(1)

Key Documents

SML3041

Sigma-Aldrich

CRBN-6-5-5-VHL

≥98% (HPLC)

Synonym(s):

CRBN-6-5-5-VHL, (2S,4R)-1-((2S)-2-(5-((5-((6-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)hexyl)oxy)pentyl)oxy)pentanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide, CRBN degrader, CRBN directed PROTAC, CRBN-VHL hetero-PROTAC

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$507.00
25 MG
$2,030.00

$507.00


Estimated to ship onApril 09, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$507.00
25 MG
$2,030.00

About This Item

Empirical Formula (Hill Notation):
C51H69N7O10S
CAS Number:
Molecular Weight:
972.20
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$507.00


Estimated to ship onApril 09, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1C2=C(C=CC=C2C(N1C3C(NC(CC3)=O)=O)=O)NCCCCCCOCCCCCOCCCCC(N[C@H](C(N4[C@H](C(NCC5=CC=C(C=C5)C6=C(C)N=CS6)=O)C[C@@H](O)C4)=O)C(C)(C)C)=O

InChI

1S/C51H69N7O10S/c1-33-44(69-32-54-33)35-20-18-34(19-21-35)30-53-46(62)40-29-36(59)31-57(40)50(66)45(51(2,3)4)55-41(60)17-8-13-28-68-27-12-7-11-26-67-25-10-6-5-9-24-52-38-16-14-15-37-43(38)49(65)58(48(37)64)39-22-23-42(61)56-47(39)63/h14-16,18-21,32,36,39-

InChI key

LIMCHOLAKDUZDS-XARBDFOFSA-N

Biochem/physiol Actions

CRBN-6-5-5-VHL is a cell-permeable and highly potent heterodimerizer comprising E3 ligases recruiting ligands, namely, pomalidomide and VH032-amine that efficiently knocks down Cereblon (CRBN). CRBN-6-5-5-VHL readily forms heteroternary complex and selectively induces CRBN ubiquitination and proteasomal degradation over VHL in a time- and dose-dependent fashion (0.001 to 1 μM). Higher concentrations (10 μM) causes the degradation of neo-substrates IKZF1 and IKZF3 with no effect on pVHL30 or pVHL19 levels.
cell-permeable and highly potent heterodimerizer comprising E3 ligases recruiting ligands, pomalidomide and VH032-amine that efficiently knocks down Cereblon (CRBN)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Steinebach et al.
MedChemComm, 10(6), 1037-1041 (2019-07-16)
A modular chemistry toolbox was developed for cereblon-directed PROTACs. A variety of linkers was attached to a CRBN ligand via the 4-amino position of pomalidomide. We used linkers of different constitution to modulate physicochemical properties. We equipped one terminus of
Christian Steinebach et al.
Chemical communications (Cambridge, England), 55(12), 1821-1824 (2019-01-24)
Small-molecule heterobifunctional degraders can effectively control protein levels and are useful research tools. We assembled proteolysis targeting chimeras (PROTACs) from a cereblon (CRBN) and a von-Hippel-Lindau (VHL) ligase ligand and demonstrated a PROTAC-induced heterodimerization of the two E3 ligases leading
Saeko Kuwahara-Ota et al.
British journal of haematology, 191(5), 784-795 (2020-06-20)
An increase in immunosuppressive myeloid-derived suppressor cells (MDSCs) is associated with disease progression and treatment resistance in multiple myeloma (MM). We investigated the mechanisms underlying MDSC induction, and sought to discover a strategy for prevention of MDSC induction in MM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service