Merck
PROTAC-mediated crosstalk between E3 ligases.
Chemical communications (Cambridge, England) (2019-01-24)
Christian Steinebach, Hannes Kehm, Stefanie Lindner, Lan Phuong Vu, Simon Köpff, Álvaro López Mármol, Corinna Weiler, Karl G Wagner, Michaela Reichenzeller, Jan Krönke, Michael Gütschow
ABSTRACT
Small-molecule heterobifunctional degraders can effectively control protein levels and are useful research tools. We assembled proteolysis targeting chimeras (PROTACs) from a cereblon (CRBN) and a von-Hippel-Lindau (VHL) ligase ligand and demonstrated a PROTAC-induced heterodimerization of the two E3 ligases leading to unidirectional and efficient degradation of CRBN.
Sigma-Aldrich
CRBN-6-5-5-VHL, ≥98% (HPLC)