Skip to Content
Merck
All Photos(1)

Key Documents

SML3029

Sigma-Aldrich

HG-9-91-01

>95% (HPLC)

Synonym(s):

1-(2,4-Dimethoxyphenyl)-3-(2,6-dimethylphenyl)-1-(6-(4-(4-methylpiperazin-1-yl)phenylamino)pyrimidin-4-yl)urea, HG 9-91-01, N-(2,4-Dimethoxyphenyl)-N′-(2,6-dimethylphenyl)-N-[6-[[4-(4-methyl-1-piperazinyl)phenyl]amino]-4-pyrimidinyl]urea

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$211.00
25 MG
$682.00

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$211.00
25 MG
$682.00

About This Item

Empirical Formula (Hill Notation):
C32H37N7O3
CAS Number:
Molecular Weight:
567.68
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

>95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=C(C)C=CC=C1C)N(C2=CC=C(OC)C=C2OC)C3=CC(NC4=CC=C(N5CCN(C)CC5)C=C4)=NC=N3

InChI

1S/C32H37N7O3/c1-22-7-6-8-23(2)31(22)36-32(40)39(27-14-13-26(41-4)19-28(27)42-5)30-20-29(33-21-34-30)35-24-9-11-25(12-10-24)38-17-15-37(3)16-18-38/h6-14,19-21H,15-18H2,1-5H3,(H,36,40)(H,33,34,35)

InChI key

UYUHRKLITDJEHB-UHFFFAOYSA-N

Biochem/physiol Actions

HG-9-91-01 is an ATP site-targeting WH-4-023 (KIN112) structural analog that inhibits salt-inducible kinases (SIKs) with improved potency (IC50 = 0.6/6.6/9.6 nM against SIK1/2/3) and selectivity, exhibiting significant inhibitory activity only against 4 kinases among a panel of 108 (% inhibition at 100 nM ∼85/EPHA2, 88/EPHA4, 91/Src, 88/YES1) and much reduced or little potency toward the others (by <67% inhibition at 100 nM), including AMPK-related kinases BTK, MARK and NUAK. HG-9-91-01 (500 nM) enhances LPS-induced IL-10 production, while suppresses proinflammatory cytokine secretion from murine marrow-derived macrophages & dendritic cells (BMDMs & BMDCs) as well as primary human macrophages in a CRTC3-dependent manner. A useful tool for studying SIKs-mediated cellular signaling events.
Potent and selective inhibitor against salt-inducible kinases SIK1, SIK2, and SIK3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service