Skip to Content
Merck
All Photos(1)

Key Documents

SML1358

Sigma-Aldrich

SGI-1027

≥98% (HPLC)

Synonym(s):

N-(4-(2-Amino-6-methylpyrimidin-4-ylamino)phenyl)-4-(quinolin-4-ylamino) benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$184.00
25 MG
$726.00

$184.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$184.00
25 MG
$726.00

About This Item

Empirical Formula (Hill Notation):
C27H23N7O
CAS Number:
Molecular Weight:
461.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$184.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(NC2=NC(N)=NC(C)=C2)C=C1)C(C=C3)=CC=C3NC4=C(C=CC=C5)C5=NC=C4

InChI

1S/C27H23N7O/c1-17-16-25(34-27(28)30-17)32-20-10-12-21(13-11-20)33-26(35)18-6-8-19(9-7-18)31-24-14-15-29-23-5-3-2-4-22(23)24/h2-16H,1H3,(H,29,31)(H,33,35)(H3,28,30,32,34)

InChI key

QSYLKMKIVWJAAK-UHFFFAOYSA-N

Application

SGI-1027 has been used as a DNA methyltransferase inhibitor to study its effects on gene expression during osteogenic differentiation.[1] It has also been used as a DNA methyltransferase 3a inhibitor to study its effects on interleukin-6 (IL-6)-induced loss of forkhead box P3 (Foxp3) gene expression in B-lymphocyte-induced maturation protein 1 (Blimp1)-Treg cells.[2]

Biochem/physiol Actions

SGI-1027 is a DNA methyltransferase (DNMT) inhibitor with IC50 values of 6-13 μM for DNMT3B, DNMT3A and DNMT1. SGI-1027 directly inhibits DNMT activity by competing with the cofactor, S-adenosylmethionine (SAM) in the methylation reaction. SGI-1027 treatment of cancer cell lines induced degradation of DNMT1, but not DNMT3A or DNMT3B, and in RKO cells caused re-expression of the silenced tumor supressor genes p16, MLH1 and TIMP3.
SGI-1027 is a DNA methyltransferase (DNMT) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Garima Garg et al.
Cell reports, 26(7), 1854-1868 (2019-02-14)
Foxp3+ regulatory T (Treg) cells restrict immune pathology in inflamed tissues; however, an inflammatory environment presents a threat to Treg cell identity and function. Here, we establish a transcriptional signature of central nervous system (CNS) Treg cells that accumulate during
Rodrigo A da Silva et al.
Bone, 125, 74-86 (2019-05-06)
The HOXA gene cluster is generally recognized as a pivotal mediator of positional identity in the skeletal system, expression of different orthologues conferring alternative locational phenotype of the vertebrate bone. Strikingly, however, the molecular mechanisms that regulate orthologue-specific expression of

Questions

  1. What is its stability at 2-8 C

    1 answer
    1. This product, when supplied in an unopened vial and stored at 2-8°C, should be stable for 1 year. If stored appropriately as supplied, it is guaranteed for 1 year from the date of shipment.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service