Skip to Content
Merck
All Photos(1)

Key Documents

SML3006

Sigma-Aldrich

ICCB-19 Hydrochloride

≥98% (HPLC)

Synonym(s):

N-Cycloheptyl-2-[(4,5-dihydro-1H-imidazol-2-yl)thio]acetamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$94.10
25 MG
$382.00

$94.10


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$94.10
25 MG
$382.00

About This Item

Empirical Formula (Hill Notation):
C12H21N3OS · HCl
CAS Number:
Molecular Weight:
291.84
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$94.10


Estimated to ship onMay 25, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1CCCCCC1)CSC2=NCCN2.Cl

InChI key

TZICOTBZKPHMMK-UHFFFAOYSA-N

Biochem/physiol Actions

ICCB-19 is a cell penetrant apoptosis inhibitor that activates autophagy leading to restoration of cellular homeostasis in cells with accumulated mutant tau, α-synuclein, or huntingtin. ICCB-19 potently and selectively inhibits TRADD. It binds to the N-terminal TRAF2-binding domain of TRADD (TRADD-N). Apparently ICCB-19 releases cIAP1/2 or TRAF2 from TRADD thus promoting the K63-linked ubiquitination of beclin 1.
cell penetrant apoptosis inhibitor that activates autophagy that potently and selectively inhibits TRADD

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daichao Xu et al.
Nature, 587(7832), 133-138 (2020-09-25)
Cell death in human diseases is often a consequence of disrupted cellular homeostasis. If cell death is prevented without restoring cellular homeostasis, it may lead to a persistent dysfunctional and pathological state. Although mechanisms of cell death have been thoroughly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service