Skip to Content
Merck
All Photos(3)

Key Documents

SML0425

Sigma-Aldrich

Caspofungin diacetate

≥97% (HPLC), powder, cell wall synthesis inhibitor

Synonym(s):

1-[(4R,5S)-5-[(2-Aminoethyl)amino]-N2-(10,12-dimethyl-1-oxotetradecyl)-4-hydroxy-L-ornithine]-5-[(3R)-3-hydroxy-L-ornithine]-pneumocandin B0

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$284.00
25 MG
$566.00

$284.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$284.00
25 MG
$566.00

About This Item

Empirical Formula (Hill Notation):
C52H88N10O15 · 2C2H4O2
CAS Number:
Molecular Weight:
1213.42
UNSPSC Code:
51283107
PubChem Substance ID:
NACRES:
NA.77

$284.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Product Name

Caspofungin diacetate, ≥97% (HPLC)

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

H2O: 15 mg/mL (clear solution)

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC([O-])=O.CC([O-])=O.CC[C@H](C)C[C@H](C)CCCCCCCCC(=O)N[C@H]1C[C@@H](O)[C@@H](NCC[NH3+])NC(=O)[C@@H]2[C@@H](O)CCN2C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H]3C[C@@H](O)CN3C(=O)[C@@H](NC1=O)[C@@H](C)O)[C@H](O)[C@@H](O)c4ccc(O)cc4)[C@H](O)CC[NH3+]

InChI

1S/C52H88N10O15.2C2H4O2/c1-5-28(2)24-29(3)12-10-8-6-7-9-11-13-39(69)56-34-26-38(68)46(55-22-21-54)60-50(75)43-37(67)19-23-61(43)52(77)41(36(66)18-20-53)58-49(74)42(45(71)44(70)31-14-16-32(64)17-15-31)59-48(73)35-25-33(65)27-62(35)51(76)40(30(4)63)57-47(34)72;2*1-2(3)4/h14-17,28-30,33-38,40-46,55,63-68,70-71H,5-13,18-27,53-54H2,1-4H3,(H,56,69)(H,57,72)(H,58,74)(H,59,73)(H,60,75);2*1H3,(H,3,4)/t28-,29+,30+,33+,34-,35-,36+,37-,38+,40-,41-,42-,43-,44-,45-,46-;;/m0../s1

InChI key

OGUJBRYAAJYXQP-IJFZAWIJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Caspofungin belongs to the echinocandins class of drugs. This drug has a limited α-phase half-life (t 1/2) of one to two hours (volume of distribution of 9.7L) and a β-phase t 1/2 of nine to 11 hours.

Application

Caspofungin diacetate has been used:

  • in an antifungal susceptibility test
  • as a standard antifungal for valproic acid (VPA)-synergy assessment
  • to treat Cryptococcus neoformans strain B3501 to study natural killer (NK) cell killing of Cryptococcus in the presence of β-1,3-glucan[1]

Biochem/physiol Actions

Caspofungin is an echinocandin antifungal that inhibits 1,3-β-D-glucan synthase which is required for cell wall synthesis.
Caspofungin is an echinocandin antifungal that inhibits 1,3-β-D-glucan synthase, which is required for cell wall synthesis, thereby disturbing the integrity of the fungal cell wall.
Caspofungin, aechinocandin, a lipophilic antifungal drug, and α β-1,3-D-glucan synthesis antagonist, are used in fungal cell wall biosynthesis research to study the role of β-1,3-D-glucans in cell wall integrity and fungal survival under various environmental and chemical stresses.[2] Caspofungin is used to treat esophageal candidiasis, candida infections, febrile neutropenia, invasive candidiasis, and invasive aspergillosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph Meletiadis et al.
Antimicrobial agents and chemotherapy, 51(6), 2053-2064 (2007-03-28)
Triple antifungal combinations are used against refractory invasive aspergillosis without an adequate understanding of their pharmacodynamic interactions. We initially studied the in vitro triple combination of voriconazole, amphotericin B, and caspofungin against Aspergillus fumigatus, A. flavus, and A. terreus by
Sarah R Beattie et al.
mSphere, 7(3), e0016322-e0016322 (2022-05-05)
There is an urgent need for new antifungals to treat cryptococcal meningoencephalitis, a leading cause of mortality in people living with HIV/AIDS. An important aspect of antifungal drug development is the validation of targets to determine whether they are required
Timothy Cairns et al.
STAR protocols, 3(4), 101883-101883 (2023-01-04)
This protocol describes procedures for quantifying Aspergillus niger growth in both solid and liquid culture. Firstly, by comparing radial growth between mutant and progenitor isolates on solid agar supplemented with sublethal stressors, susceptibility coefficients can be calculated. Secondly, analysis of
Liping Xiong et al.
mSphere, 9(7), e0037224-e0037224 (2024-07-09)
Iron acquisition is critical for pathogens to proliferate during invasive infection, and the human fungal pathogen Candida albicans is no exception. The iron regulatory network, established in reference strain SC5314 and derivatives, includes the central player Sef1, a transcription factor
Esther Sheba et al.
The American journal of tropical medicine and hygiene, 108(2), 377-383 (2022-12-27)
This study describes the microbiological and histopathological features of patients with COVID-19-associated rhino-orbital mucormycosis (ROM) seen at the L V Prasad Eye Institute between May and August 2021. Diagnosed clinically and radiologically, 24 patients with ROM were included in the

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service