Skip to Content
Merck
All Photos(1)

Key Documents

SML2993

Sigma-Aldrich

JH-RE-06

≥98% (HPLC)

Synonym(s):

8-Chloro-2-[(2,4-dichlorophenyl)amino]-3-(3-methyl-1-oxobutyl)-5-nitro-4(1H)-quinolinone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$150.00
25 MG
$596.00

$150.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$150.00
25 MG
$596.00

About This Item

Empirical Formula (Hill Notation):
C20H16Cl3N3O4
CAS Number:
Molecular Weight:
468.72
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$150.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1c(ccc(c1)Cl)Nc2[nH]c3c([c](c2C(=O)CC(C)C)=O)c(ccc3Cl)[N+](=O)[O-]

InChI

1S/C20H16Cl3N3O4/c1-9(2)7-15(27)17-19(28)16-14(26(29)30)6-4-11(22)18(16)25-20(17)24-13-5-3-10(21)8-12(13)23/h3-6,8-9H,7H2,1-2H3,(H2,24,25,28)

InChI key

LRTXIQCBQIKIOH-UHFFFAOYSA-N

Biochem/physiol Actions

JH-RE-06 is a potent and selective inhibitor of REV1-REV7 interaction that induces REV1 dimerization blocking the REV1-REV7 interaction and POL ϲ recruitment. JH-RE-06 potently inhibits translesion synthesis (TLS) and augments cisplatin effectiveness in cultured human and mouse cell lines. It improves effectiveness of cisplatin in A375 tumor xenograft mouse model.
potent and selective inhibitor of REV1-REV7 interaction blocking the REV1-REV7 interaction and POL ϲ recruitment

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kavi P M Mehta et al.
Cell reports, 31(9), 107705-107705 (2020-06-04)
5-Hydroxymethylcytosine (5hmC) binding, ES-cell-specific (HMCES) crosslinks to apurinic or apyrimidinic (AP, abasic) sites in single-strand DNA (ssDNA). To determine whether HMCES responds to the ssDNA abasic site in cells, we exploited the activity of apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like
Jessica L Wojtaszek et al.
Cell, 178(1), 152-159 (2019-06-11)
Intrinsic and acquired drug resistance and induction of secondary malignancies limit successful chemotherapy. Because mutagenic translesion synthesis (TLS) contributes to chemoresistance as well as treatment-induced mutations, targeting TLS is an attractive avenue for improving chemotherapeutics. However, development of small molecules with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service