Skip to Content
Merck
All Photos(1)

Key Documents

SML2992

Sigma-Aldrich

uPSEM792 hydrochloride

≥98% (HPLC)

Synonym(s):

1-Methyl-7,8,9,10-tetrahydro-1H-6,10-methanoazepino[4,5-g]quinoxalin-2(6H)-one hydrochloride, PSEM 792 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$225.00
25 MG
$895.00

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$225.00
25 MG
$895.00

About This Item

Empirical Formula (Hill Notation):
C14H15N3O · HCl
CAS Number:
Molecular Weight:
277.75
UNSPSC Code:
12352200
NACRES:
NA.77

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N(C)C2=CC(C3CNCC4C3)=C4C=C2N=C1.Cl

InChI key

CDHPEJUYEXNGCV-UHFFFAOYSA-N

Biochem/physiol Actions

Ultrapotent PSEM agonist for chimeric ligand-gated ion channel pores with nAChR α7 LBD L181G/Q139L/Y217F mutant (PSAM4) in vitro and in vivo.
uPSEM792 is a selective and ultrapotent pharmacologically selective effector molecule (uPSEM) agonist for chimeric pharmacologically selective actuator modules (PSAMs) with mutant nAChR α7 ligand-binding domain (LBD) PSML181G/Q139L/Y217F (PSAM4) fused to the ion pore domain (IPD) of ligand-gated ion channels (LGICs), including glycine receptor (PSAM4-GlyR EC50 = 2.3 nM) and 5-HT3 (PSAM4-5-HT3 EC50 = 1.6 nM). uPSEM792 exhibits high PSM4 selectivity (>10,000-fold over α7-GlyR, α7-5HT3, 5HT3; >230-fold over α4β2 nAChR) and sub-nanomolar PSAM4-GlyR affinity (Ki = 0.7 nM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Magnus et al.
Science (New York, N.Y.), 364(6436) (2019-03-16)
Chemogenetics enables noninvasive chemical control over cell populations in behaving animals. However, existing small-molecule agonists show insufficient potency or selectivity. There is also a need for chemogenetic systems compatible with both research and human therapeutic applications. We developed a new

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service