Skip to Content
Merck
All Photos(1)

Key Documents

SML2944

Sigma-Aldrich

Mca-YVADAPK(Dnp)-OH trifluoroacetate

≥97% (HPLC)

Synonym(s):

Mca-Tyr-Val-Ala-Asp-Ala-Pro-Lys(Dnp), TFA salt, Mca-Tyr-Val-Ala-Asp-Ala-Pro-Lys(Dnp)-OH, TFA salt, Mca-YVADAPK(Dnp), TFA salt (Mca = 7-Methoxycoumarin-4-yl; Dnp = 2,4-dinitrophenyl)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$301.00

$301.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$301.00

About This Item

Empirical Formula (Hill Notation):
C53H64N10O19 · xC2HF3O2
Molecular Weight:
1145.13 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$301.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

film or powder

color

white to dark beige

storage temp.

−20°C

Biochem/physiol Actions

Mca-YVADAPK(Dnp)-OH is a widely used fluorescent substrate for caspase-1 (ICE cleaves between D & A) and angiotensin-converting enzyme 2 (ACE2 cleaves between P & K). Proteolytic cleavage separates 7-methoxycoumarin-4-acetyl (Mca) from the fluorescence-quenching group 2,4-dinitrophenyl (Dnp), resulting in increased Mca fluorescence (Ex: 320-328 nm, Em: 405-420 nm).
Popular fluorescent substrate for caspase-1 (ICE) and angiotensin-converting enzyme 2 (ACE2).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin B Bruce et al.
Experimental gerontology, 111, 133-140 (2018-07-15)
The obesity epidemic is multi-generational and is particularly debilitating in the aging population, necessitating the use of pharmaceutical interventions. Recent evidence suggests that increasing the activity of the angiotensin converting enzyme-2 [ACE2]/angiotensin-(1-7)[Ang-(1-7)]/Mas receptor (MasR) axis in obese animal models leads
Jiantao Song et al.
Bioscience reports, 40(6) (2020-05-29)
Angiotensin-converting enzyme 2 (ACE2) is considered as an endogenous negative regulator of renin-angiotensin system (RAS), exerting multiple cardiovascular protective roles. Whether mechanical stretch modulates ACE2 expression remains unknown. The present study aimed at investigating whether ACE2 is involved in physiological
M Enari et al.
Nature, 380(6576), 723-726 (1996-04-25)
Binding of Fas ligand or an agonistic anti-Fas antibody induces apoptosis in Fas-bearing cells. The interleukin-1Beta-converting enzyme (ICE) is a cysteine protease that is involved in apoptosis induced by various stimuli, including Fas-mediated apoptosis. Several ICE homologues have been identified
Chhinder P Sodhi et al.
Journal of immunology (Baltimore, Md. : 1950), 203(11), 3000-3012 (2019-10-28)
Angiotensin-converting enzyme 2 (ACE2) is a potent negative regulator capable of restraining overactivation of the renin-angiotensin system, which contributes to exuberant inflammation after bacterial infection. However, the mechanism through which ACE2 modulates this inflammatory response is not well understood. Accumulating
Chad Vickers et al.
The Journal of biological chemistry, 277(17), 14838-14843 (2002-01-30)
Human angiotensin-converting enzyme-related carboxypeptidase (ACE2) is a zinc metalloprotease whose closest homolog is angiotensin I-converting enzyme. To begin to elucidate the physiological role of ACE2, ACE2 was purified, and its catalytic activity was characterized. ACE2 proteolytic activity has a pH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service