Skip to Content
Merck
All Photos(1)

Key Documents

SML2926

Sigma-Aldrich

FT895

≥98% (HPLC)

Synonym(s):

2,3-Dihydro-N-hydroxy-1,1-dimethyl-2-[5-(trifluoromethyl)-2-pyrazinyl]-1H-isoindole-4-carboxamide, FT 895, FT-895, HDTK 070, HDTK-070, HDTK070, N-Hydroxy-1,1-dimethyl-2-(5-(trifluoromethyl)pyrazin-2-yl)isoindoline-4-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$135.00
25 MG
$544.00

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$135.00
25 MG
$544.00

About This Item

Empirical Formula (Hill Notation):
C16H15F3N4O2
CAS Number:
Molecular Weight:
352.31
UNSPSC Code:
12352200
NACRES:
NA.77

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1ncc(nc1)N2C(c3c(c(ccc3)C(=O)NO)C2)(C)C

InChI key

XCLPBILTRRWOIL-UHFFFAOYSA-N

Biochem/physiol Actions

FT895 is a potent and highly selective HDAC11 inhibitor (IC50 = 3 nM/HDAC11, 5.6 μM/HDAC8, >10 μM/HDAC1-7, HDAC9 & HDAC10) with a deacetylase active site Zn2+-targeting hydroxamate. FT895 exhibits antiproliferation potency against myeloproliferative neoplasm (MPN) cultures (IC50 = 3.00 μM/JAK2(V617F) Ba/F3, 4.94 μM/HEL92.1.7, 6.49 μM/MPL(W515L) Ba/F3) and reduces stem-like properties of CSCs (42/62/85/100% inhibtion of H1650 Sphere formation at 0.5/1/2/5 μM).
Potent and highly selective HDAC11 inhibitor that reduces myeloproliferative neoplasm (MPN) proliferation and CSCs stem-like properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Se In Son et al.
ACS chemical biology, 14(7), 1393-1397 (2019-07-03)
Mammalian histone deacetylases (HDACs) are a class of enzymes that play important roles in biological pathways. Existing HDAC inhibitors target multiple HDACs without much selectivity. Inhibitors that target one particular HDAC will be useful for investigating the biological functions of
Matthew W Martin et al.
Bioorganic & medicinal chemistry letters, 28(12), 2143-2147 (2018-05-20)
N-Hydroxy-2-arylisoindoline-4-carboxamides are potent and selective inhibitors of HDAC11. The discovery, synthesis, and structure activity relationships of this novel series of inhibitors are reported. An advanced analog (FT895) displays promising cellular activity and pharmacokinetic properties that make it a useful tool
Lanzhu Yue et al.
Blood, 135(3), 191-207 (2019-11-22)
Protein acetylation is an important contributor to cancer initiation. Histone deacetylase 6 (HDAC6) controls JAK2 translation and protein stability and has been implicated in JAK2-driven diseases best exemplified by myeloproliferative neoplasms (MPNs). By using novel classes of highly selective HDAC
Namrata Bora-Singhal et al.
Scientific reports, 10(1), 4722-4722 (2020-03-15)
Non-small cell lung cancer (NSCLC) is known to have poor patient outcomes due to development of resistance to chemotherapy agents and the EGFR inhibitors, which results in recurrence of highly aggressive lung tumors. Even with recent success in immunotherapy using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service