Skip to Content
Merck
All Photos(1)

Key Documents

SML2824

Sigma-Aldrich

Valiglurax

≥98% (HPLC)

Synonym(s):

N-(1H-Pyrazolo[3,4-b]pyridin-3-yl)-1-(trifluoromethyl)isoquinolin-6-amine, N-1H-Pyrazolo[3,4-b]pyridin-3-yl-1-(trifluoromethyl)-6-isoquinolinamine, VU0652957, VU2957

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$118.00
25 MG
$475.00

$118.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$118.00
25 MG
$475.00

About This Item

Empirical Formula (Hill Notation):
C16H10F3N5
CAS Number:
Molecular Weight:
329.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$118.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1nccc2c1ccc(c2)Nc3n[nH]c4ncccc43

InChI

1S/C16H10F3N5/c17-16(18,19)13-11-4-3-10(8-9(11)5-7-20-13)22-15-12-2-1-6-21-14(12)23-24-15/h1-8H,(H2,21,22,23,24)

InChI key

RUEXKBWCUUFJMY-UHFFFAOYSA-N

Biochem/physiol Actions

Valiglurax is an orally available, CNS penetrant, potent and selective mGlu4 positive allosteric modulator (PAM).
orally available, CNS penetrant, potent and selective mGlu4 PAM

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph D Panarese et al.
ACS medicinal chemistry letters, 10(3), 255-260 (2019-03-21)
Herein, we report the discovery of a novel potent, selective, CNS penetrant, and orally bioavailable mGlu4 PAM, VU0652957 (VU2957, Valiglurax). VU2957 possessed attractive in vitro and in vivo pharmacological and DMPK properties across species. To advance toward the clinic, a
Joseph D Panarese et al.
Bioorganic & medicinal chemistry letters, 29(2), 342-346 (2018-12-07)
This letter describes the first account of the chemical optimization (SAR and DMPK profiling) of a new series of mGlu4 positive allosteric modulators (PAMs), leading to the identification of VU0652957 (VU2957, Valiglurax), a compound profiled as a preclinical development candidate.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service