Skip to Content
Merck
All Photos(1)

Key Documents

SML1145

Sigma-Aldrich

ML303

≥98% (HPLC)

Synonym(s):

2-Methoxy-4-(3-methyl-1-(4-(trifluoromethyl)phenyl)-1H-pyrazolo[3,4-b]pyridin-4-yl)phenol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$447.00

$447.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$447.00

About This Item

Empirical Formula (Hill Notation):
C21H16F3N3O2
CAS Number:
Molecular Weight:
399.37
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$447.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=NN(C2=CC=C(C(F)(F)F)C=C2)C3=NC=CC(C4=CC=C(O)C(OC)=C4)=C31

InChI

1S/C21H16F3N3O2/c1-12-19-16(13-3-8-17(28)18(11-13)29-2)9-10-25-20(19)27(26-12)15-6-4-14(5-7-15)21(22,23)24/h3-11,28H,1-2H3

InChI key

YBRHGNCGFMRGSV-UHFFFAOYSA-N

Biochem/physiol Actions

It belongs to the pyrazolopyridine family.[1]
ML303 is a potent inhibitor (IC50 = 155 nM) of the influenza viral protien NS1. NS1 enhances virulence by inhibiting host cell immune responses, including the expression of IFN-b, and regulating synthesis, splicing and translation of viral RNA.
ML303 is a potent inhibitor of the influenza viral protien NS1.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nícia Rosário-Ferreira et al.
International journal of molecular sciences, 21(4) (2020-02-27)
Influenza (flu) is a contagious viral disease, which targets the human respiratory tract and spreads throughout the world each year. Every year, influenza infects around 10% of the world population and between 290,000 and 650,000 people die from it according

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service