Skip to Content
Merck
All Photos(1)

Key Documents

SML2231

Sigma-Aldrich

Erasin

≥98% (HPLC)

Synonym(s):

(E)-N′-((6-(tert-butyl)-4-oxo-4H-chromen-3-yl)methylene)nicotinohydrazide, Erlotinib-resistance antagonizing STAT3 inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$162.00
25 MG
$653.00

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$162.00
25 MG
$653.00

About This Item

Empirical Formula (Hill Notation):
C20H19N3O3
CAS Number:
Molecular Weight:
349.38
UNSPSC Code:
12352200
NACRES:
NA.77

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C(/C=N/NC(C2=CC=CN=C2)=O)=COC3=CC=C(C(C)(C)C)C=C31

InChI

1S/C20H19N3O3/c1-20(2,3)15-6-7-17-16(9-15)18(24)14(12-26-17)11-22-23-19(25)13-5-4-8-21-10-13/h4-12H,1-3H3,(H,23,25)/b22-11+

InChI key

JHUVYXUBEMYTNO-SSDVNMTOSA-N

Biochem/physiol Actions

Erasin (Erlotinib-resistance antagonizing STAT3 inhibitor) is a selective inhibitor of STAT3 with selectivity over STAT5 and STAT1 in cell-based assays. It was shown to increase the apoptotic rate of HCC-827 cells with acquired resistance against Erlotinib in addition to increasing the apoptotic rate of cultured NSCLC cells in a STAT3-dependent manner.
Selective inhibitor of STAT3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Lis et al.
Scientific reports, 7(1), 17390-17390 (2017-12-14)
Inhibition of protein-protein interactions by small molecules offers tremendous opportunities for basic research and drug development. One of the fundamental challenges of this research field is the broad lack of available lead structures from nature. Here, we demonstrate that modifications

Questions

  1. I need a list of the percentage of ingredients in ERASI for products going to Canada. Required by health canada for personal care products.

    1 answer
    1. Please see section 3 and section 15 of the Safety Data Sheet:
      https://www.sigmaaldrich.com/sds/sigma/sml2231.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service