Skip to Content
Merck
All Photos(1)

Key Documents

SML2706

Sigma-Aldrich

Jadomycin B

≥98% (HPLC)

Synonym(s):

1-(sec-butyl)-12-((4,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-7-hydroxy-5-methyl-8H-benzo[b]oxazolo[3,2-f]phenanthridine-2,8,13(1H,3aH)-trione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H31NO9
CAS Number:
Molecular Weight:
549.57
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

Streptomyces venezuelae

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

solubility

DMSO: 1 mg/mL

storage temp.

−20°C

SMILES string

N21C(OC(=O)C2C(CC)C)c3c(c(cc(c3)C)O)C4=C1C(=O)c5c(cccc5OC6OC(C(C(C6)O)O)C)C4=O

InChI

1S/C30H31NO9/c1-5-13(3)24-30(37)40-29-16-9-12(2)10-17(32)21(16)23-25(31(24)29)28(36)22-15(27(23)35)7-6-8-19(22)39-20-11-18(33)26(34)14(4)38-20/h6-10,13-14,18,20,24,26,29,32-34H,5,11H2,1-4H3

InChI key

BSBSCJRAEMDCHC-UHFFFAOYSA-N

General description

Jadomycin B, is produced by Streptomyces venezuelae fermentation. It is an angucycline antibiotic containing an unusual 8H-benz[b]oxazolo[3,2-f]-phenanthridine ring system and a rare carbohydrate, digitoxose.1,2
Jadomycin B displays antimicrobial, anti-tumor, aurora-B kinase inhibition, DNA cleaving and more activities.3,4,5,6
Jadomycin B was found to be active against a variety of staphylococci, including methicillin-resistant Staphylococcus aureus in a MIC of 1μg/ml.3 In addition, its anti-tumor activity was demonstrated as it kills drug-sensitive and multidrug-resistant breast cancer cell, through inhibition of type II topoisomerases and the induction of DNA damage and apoptosis. Jadomycin B (15 mM), 24-hour treatment significantly lowered the levels of topoisomerase IIa protein versus the vehicle control.4
It was also shown that Jadomycin B inhibits Aurora-B kinase activity by phosphorylation of histone H3 on Ser10 in a dose-dependent manner (10μg /mL Jadomycin B reduced H3 phosphorylation by 70%).5
Jadomycin B was also found to cleave DNA in the presence of Cu (II) by reducing it to Cu(I) which can further react with H2O2 to form hydroxyl radicals that causes DNA strand scission without the requirement of any external reducing agent. The EC50 value of Jadomycin B for single-strand scission was approximately 1.7μM.6

Biochem/physiol Actions

Anti cancer, anti microbial

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jadomycin, a novel 8H-benz[b]oxazolo[3,2-f]-phenanthridine antibiotic from streptomyces venezuelae ISP5230
Ayer S. W. et al
Tetrahedron Letters, 6301-6304 (1991)
Susan M A Monro et al.
Bioorganic & medicinal chemistry, 19(11), 3357-3360 (2011-05-14)
The natural product jadomycin B, isolated from Streptomyces venezeulae ISP5230, has been found to cleave DNA in the presence of Cu(II) ions without the requirement for an external reducing agent. The efficiency of DNA cleavage was probed using supercoiled plasmid
Da-Hua Fu et al.
Molecular cancer therapeutics, 7(8), 2386-2393 (2008-08-30)
Aurora kinases have emerged as promising targets for cancer therapy because of their critical role in mitosis. These kinases are well-conserved in all eukaryotes, and IPL1 gene encodes the single Aurora kinase in budding yeast. In a virtual screening attempt
Steven R Hall et al.
The Journal of pharmacology and experimental therapeutics, 363(2), 196-210 (2017-09-15)
Jadomycins are natural products that kill drug-sensitive and multidrug-resistant (MDR) breast cancer cells. To date, the cytotoxic activity of jadomycins has never been tested in MDR breast cancer cells that are also triple negative. Additionally, there is only a rudimentary
Liru Wang et al.
Microbiology (Reading, England), 148(Pt 4), 1091-1103 (2002-04-05)
Eight additional genes, jadX, O, P, Q, S, T, U and V, in the jad cluster of Streptomyces venezuelae ISP5230, were located immediately downstream of jadN by chromosome walking. Sequence analyses and comparisons implicated them in biosynthesis of the 2,6-dideoxysugar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service