Skip to Content
Merck
All Photos(1)

Key Documents

SML2624

Sigma-Aldrich

Dexlansoprazole

≥98% (HPLC)

Synonym(s):

(R)-(+)-Lansoprazole, (R)-(+)2-([3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl)-1H-benzo[d]imidazole

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$148.00

$148.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$148.00

About This Item

Empirical Formula (Hill Notation):
C16H14F3N3O2S
CAS Number:
Molecular Weight:
369.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$148.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +142 to +158°, c = 1 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C16H14F3N3O2S/c1-10-13(20-7-6-14(10)24-9-16(17,18)19)8-25(23)15-21-11-4-2-3-5-12(11)22-15/h2-7H,8-9H2,1H3,(H,21,22)/t25-/m1/s1

InChI key

MJIHNNLFOKEZEW-RUZDIDTESA-N

Biochem/physiol Actions

Dexlansoprazole is a proton pump inhibitor. It is the active enantiomer of lansoprazole used to treat gastroesophageal reflux disease (GERD).
Proton pump inhibitor; Active enantiomer of lansoprazole to treat gastroesophageal reflux disease (GERD)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khean Lee Goh et al.
Journal of neurogastroenterology and motility, 22(3), 355-366 (2016-03-05)
Although gastroesophageal reflux disease is not as common in Asia as in western countries, the prevalence has increased substantially during the past decade. Gastroesophageal reflux disease is associated with considerable reductions in subjective well-being and work productivity, as well as
Karen E Hansen et al.
Gastroenterology, 156(4), 926-934 (2018-11-18)
Epidemiological studies have associated proton pump inhibitor (PPI) therapy with osteoporotic fractures, but it is not clear if PPIs directly cause osteoporosis. We evaluated the effect of dexlansoprazole and esomeprazole on bone turnover, bone mineral density (BMD), true fractional calcium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service