Skip to Content
Merck
All Photos(1)

Documents

SML3032

Sigma-Aldrich

Eletriptan hydrobromide

≥98% (HPLC)

Synonym(s):

3-[[(2R)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-indole hydrobromide, UK-116,044, UK-116044

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O2S · HBr
CAS Number:
Molecular Weight:
463.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

Br.CN1CCCC1Cc2c[nH]c3ccc(CCS(=O)(=O)c4ccccc4)cc23

InChI

1S/C22H26N2O2S.BrH/c1-24-12-5-6-19(24)15-18-16-23-22-10-9-17(14-21(18)22)11-13-27(25,26)20-7-3-2-4-8-20;/h2-4,7-10,14,16,19,23H,5-6,11-13,15H2,1H3;1H/t19-;/m1./s1

InChI key

UTINOWOSWSPFLJ-FSRHSHDFSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Eletriptan hydrobromide is a serotonin 5-HT1B/1D receptor agonist; second generation anti-migraine drug.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Willems et al.
Naunyn-Schmiedeberg's archives of pharmacology, 358(2), 212-219 (1998-09-28)
It has been suggested that opening of cephalic arteriovenous anastomoses may be involved in the headache phase of migraine. Indeed, a number of acutely acting anti-migraine drugs, including the ergot alkaloids and sumatriptan, constrict porcine carotid arteriovenous anastomoses. In this
C Napier et al.
European journal of pharmacology, 368(2-3), 259-268 (1999-04-08)
The affinity of eletriptan ((R)-3-(1-methyl-2-pyrrolidinylmethyl)-5-[2-(phenylsulphonyl )ethyl]-1H-indole) for a range of 5-HT receptors was compared to values obtained for other 5-HT1B/1D receptor agonists known to be effective in the treatment of migraine. Eletriptan, like sumatriptan, zolmitriptan, naratriptan and rizatriptan had highest
P Gupta et al.
European journal of pharmacology, 367(2-3), 283-290 (1999-03-17)
The functional activity of eletriptan ((R)-3-(1-methyl-2-pyrrolidinylmethyl)-5-[2-(phenylsulphonyl )ethyl]- 1 H-indole) at the contractile serotonin (5-hydroxytryptamine; 5-HT) '1B-like' receptor in dog isolated saphenous vein and basilar artery was investigated. Eletriptan, like 5-HT and sumatriptan potently contracted saphenous vein (pEC50: 6.3, 6.9 and
L A Dostal et al.
Teratology, 50(6), 387-394 (1994-12-01)
The developmental toxicity of the 3-hydroxy-3-methylglutaryl-coenzyme A (HMG-CoA) reductase inhibitor, atorvastatin, was investigated in pregnant rats and rabbits given daily oral doses during organogenesis. Rats received 0, 10, 100, or 300 mg/kg on days 6-15 of gestation, and rabbits received

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service