Skip to Content
Merck
All Photos(1)

Key Documents

SML2611

Sigma-Aldrich

Bobcat339

≥98% (HPLC)

Synonym(s):

1-([1,1′-Biphenyl]-3-yl)-4-amino-5-chloropyrimidin-2(1H)-one, 4-Amino-1-[1,1′-biphenyl]-3-yl-5-chloro-2(1H)-pyrimidinone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$146.00
25 MG
$590.00

$146.00


Estimated to ship onApril 21, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$146.00
25 MG
$590.00

About This Item

Empirical Formula (Hill Notation):
C16H12ClN3O
CAS Number:
Molecular Weight:
297.74
UNSPSC Code:
12352200
NACRES:
NA.77

$146.00


Estimated to ship onApril 21, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=NC(N(C2=CC=CC(C3=CC=CC=C3)=C2)C=C1Cl)=O

InChI

1S/C16H12ClN3O/c17-14-10-20(16(21)19-15(14)18)13-8-4-7-12(9-13)11-5-2-1-3-6-11/h1-10H,(H2,18,19,21)

InChI key

QMGYGOOYCNTEQO-UHFFFAOYSA-N

Biochem/physiol Actions

Bobcat339 is a cell penetrant, potent and selective inhibitor of TET1 and TET2 [Ten-eleven translocation methylcytosine dioxygenase (TET) 1 and 2] that reduces content of 5-hydroxymethyl-cytosine (5hmC) in cultured neurons. Bobcat339 does not inhibit DNMT3a.
cell penetrant, potent and selective inhibitor of TET1 and TET2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriella N L Chua et al.
ACS medicinal chemistry letters, 10(2), 180-185 (2019-02-21)
DNA methylation is known as the prima donna epigenetic mark for its critical role in regulating local gene transcription. Changes in the landscape of DNA methylation across the genome occur during cellular transition, such as differentiation and altered neuronal plasticity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service