Skip to Content
Merck
All Photos(1)

Key Documents

SML1972

Sigma-Aldrich

LLY-283

≥98% (HPLC)

Synonym(s):

(2R,3R,4S,5R)-2-(4-Amino-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-5-((R)-hydroxy(phenyl)methyl)tetrahydrofuran-3,4-diol, (R)-5′-phenyl-7-deazaadenosine; 6-amino-9-[(R)-5′-phenyl(ribofuranosyl)]-7-deazapurine, LLY 283, LLY283

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$281.00
25 MG
$706.00

$281.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$281.00
25 MG
$706.00

About This Item

Empirical Formula (Hill Notation):
C17H18N4O4
CAS Number:
Molecular Weight:
342.35
UNSPSC Code:
12352200
NACRES:
NA.77

$281.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H]([C@H](O)C2=CC=CC=C2)O[C@@H](N3C(N=CN=C4N)=C4C=C3)[C@@H]1O

InChI key

WWOOWAHTEXIWBO-QFRSUPTLSA-N

Biochem/physiol Actions

A potent and selective SAM-competitive PRMT5 inhibitor.
LLY-283 is a potent and selective SAM-competitive PRMT5 inhibitor (IC50 = 20 nM against PRMT5-catalyzed methylation of an H4R3-derived peptide substrate) with greater than 100-fold selectivity over other histone methyltransferases and non-epigenetic targets. LLY-283 inhibits Smith (Sm) antigen SmBB′ methylation in MCF7 cells (IC50 = 25 nM; 3 days) and affects MDM4 splicing in A375 cells (IC50 = 40 nM; 3 days). The diastereomer LLY-284 is less potent in the biochemical assay and is an ideal control compound for cellular studies. For characterization details of LLY-283, please visit the LLY-283 probe summary on the Structural Genomics Consortium (SGC) website.

LLY-284 is the negative control for the active probe, LLY-283. LLY-284 is available from Sigma. To learn more about and purchase LLY-284, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

LLY-283 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service