Skip to Content
Merck
All Photos(1)

Key Documents

SML2567

Sigma-Aldrich

LY2784544

≥98% (HPLC)

Synonym(s):

3-(4-Chloro-2-fluorobenzyl)-2-methyl-N-(5-methyl-1H-pyrazol-3-yl)-8-(morpholinomethyl)imidazo[1,2-b]pyridazin-6-amine, 3-[(4-Chloro-2-fluorophenyl)methyl]-2-methyl-N-(5-methyl-1H-pyrazol-3-yl)-8-(4-morpholinylmethyl)imidazo[1,2-b]pyridazin-6-amine, LY 2784544, LY-2784544

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$158.00
25 MG
$507.00

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$158.00
25 MG
$507.00

About This Item

Empirical Formula (Hill Notation):
C23H25ClFN7O
CAS Number:
Molecular Weight:
469.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

composition

H2O, 0-4 mol/mol (moles per mole of product)

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(ccc(c1)Cl)Cc2[n]3c(nc2C)C(=CC(=N3)Nc5n[nH]c(c5)C)CN4CCOCC4

InChI

1S/C23H25ClFN7O/c1-14-9-21(29-28-14)27-22-11-17(13-31-5-7-33-8-6-31)23-26-15(2)20(32(23)30-22)10-16-3-4-18(24)12-19(16)25/h3-4,9,11-12H,5-8,10,13H2,1-2H3,(H2,27,28,29,30)

InChI key

SQSZANZGUXWJEA-UHFFFAOYSA-N

Biochem/physiol Actions

LY2784544 is an orally active, ATP-competitive, potent and JAK2-selective janus tyrosine kinase inhibitor (IC50/[ATP] = 2.52 nM/5 μM/JAK2, 19.8 nM/20 μM/JAK1, 48.0 nM/2 μM/JAK3) with little activity toward 79 other kinases. LY2784544 selectively inhibits signaling/proliferation driven by oncogenic JAK2V617F (IC50 = 20/55 nM) over those mediated by wt JAK2 upon IL-3 induction (IC50 = 1183/1309 nM) in Ba/F3 cultures and displays in vivo efficacious in a JAK2V617F-induced murine MPN model (10-80 mg/kg bid po.). LY2784544 is also reported to display Zn-dependent (optimal [Zn+2] = 100 μM) GPR39 agonist activity, but not 11 other GPRs.
Orally active, ATP-competitive, potent and JAK2-selective janus tyrosine kinase inhibitor with additional Zn+2-dependent GPR39 agonist activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul J Guyett et al.
ACS infectious diseases, 2(7), 518-528 (2016-09-15)
Human parasite Trypanosoma brucei proliferates in the blood of its host, where it takes up iron via receptor-mediated endocytosis of transferrin (Tf). Mechanisms of Tf endocytosis in the trypanosome are not fully understood. Small molecule lapatinib inhibits Tf endocytosis in
Wei Zhou et al.
Oncogene, 37(29), 3981-3997 (2018-04-18)
Antineoplastic platinum agents are used in first-line treatment of ovarian cancer, but treatment failure frequently results from platinum drug resistance. Emerging observations suggest a role of reactive oxygen species (ROS) in the resistance of cancer drugs including platinum drugs. However
L Ma et al.
Blood cancer journal, 3, e109-e109 (2013-04-16)
Owing to the prevalence of the JAK2V617F mutation in myeloproliferative neoplasms (MPNs), its constitutive activity, and ability to recapitulate the MPN phenotype in mouse models, JAK2V617F kinase is an attractive therapeutic target. We report the discovery and initial characterization of
Nuramalina H Mumin et al.
BMC cancer, 19(1), 102-102 (2019-01-27)
Due to the lack of effective therapies and poor prognosis in TNBC (triple-negative breast cancer) patients, there is a strong need to develop effective novel targeted therapies for this subtype of breast cancer. Inhibition of heat shock protein 90 (HSP90)
Qian Sun et al.
Biochemical and biophysical research communications, 498(1), 164-170 (2018-02-07)
Apigetrin (APG), as a flavonoid, has many cellular bioactivities, including regulation of oxidative stress, and induction of apoptosis. However, the means by which APG suppresses human gastric cancer are still little to be understood. In the present study, the anti-cancer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service