Skip to Content
Merck
All Photos(1)

Key Documents

SML2558

Sigma-Aldrich

N-arachidonoylglycine

≥98% (HPLC)

Synonym(s):

N-[(5Z,8Z,11Z,14Z)-1-oxo-5,8,11,14-eicosatetraen-1-yl]-glycine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$61.00
25 MG
$247.00

$61.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$61.00
25 MG
$247.00

About This Item

Empirical Formula (Hill Notation):
C22H35NO3
CAS Number:
Molecular Weight:
361.52
UNSPSC Code:
12352211
NACRES:
NA.77

$61.00


Estimated to ship onMay 13, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)NCC(O)=O

InChI

1S/C22H35NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-21(24)23-20-22(25)26/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-20H2,1H3,(H,23,24)(H,25,26)/b7-6-,10-9-,13-12-,16-15-

InChI key

YLEARPUNMCCKMP-DOFZRALJSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Arachidonylglycine (NAGly) is a metabolite of the endocannabinoid anandamide (AEA) that binds to G-protein coupled receptor 18 (GPR18). It is an endogenous inhibitor of fatty acid amide hydrolase (FAAH), and also shown to be an inhibitor of the glycine transporter GlyT2. NAGly is involved in a number of pathways associated with analgesic activity and has exhibited analgesic properties in a rat model of inflammatory pain

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service