N-arachidonoylglycine
Empirical Formula (Hill Notation):
C22H35NO3
CAS Number:
Molecular Weight:
361.52
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Research Area
Showing 1-2 of 2
Compare
Product Number
Description
Pricing