Skip to Content
Merck
All Photos(1)

Key Documents

SML2519

Sigma-Aldrich

Anamorelin

≥98% (HPLC)

Synonym(s):

2-Amino-N-[(2R)-1-[(3R)-3-benzyl-3-[dimethylamino(methyl)carbamoyl]piperidin-1-yl]-3-(1H-indol-3-yl)-1-oxopropan-2-yl]-2-methylpropanamide, ONO-7643, RC-1291, ST-1291

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$109.00
25 MG
$443.00

$109.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$109.00
25 MG
$443.00

About This Item

Empirical Formula (Hill Notation):
C31H42N6O3
CAS Number:
Molecular Weight:
546.70
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$109.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

N3(C[C@@](CCC3)(Cc4ccccc4)C(=O)N(N(C)C)C)C(=O)[C@H](NC(=O)C(N)(C)C)Cc1c2c([nH]c1)cccc2

InChI

1S/C31H42N6O3/c1-30(2,32)28(39)34-26(18-23-20-33-25-15-10-9-14-24(23)25)27(38)37-17-11-16-31(21-37,29(40)36(5)35(3)4)19-22-12-7-6-8-13-22/h6-10,12-15,20,26,33H,11,16-19,21,32H2,1-5H3,(H,34,39)/t26-,31-/m1/s1

InChI key

VQPFSIRUEPQQPP-MXBOTTGLSA-N

Biochem/physiol Actions

Anamorelin is a non-peptidic ghrelin mimetic and growth hormone (GH) secretagogue. It is an orally-active, brain-penetrant, selective agonist of the ghrelin/growth hormone secretagogue receptor (GHSR). Anamorelin has been shown to increases appetite, overall body weight, lean body mass, and muscle strength in clinical studies with cancer patients suffering from cachexia.
Ghrelin receptor agonist

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuyuki Katakami et al.
Cancer, 124(3), 606-616 (2017-12-06)
Cachexia, described as weight loss (mainly in lean body mass [LBM]) and anorexia, is common in patients with advanced cancer. This study examined the efficacy and safety of anamorelin (ONO-7643), a novel selective ghrelin receptor agonist, in Japanese cancer patients
Palliative care: Anamorelin provides benefit to patients with cachexia.
Peter Sidaway
Nature reviews. Clinical oncology, 15(2), 68-68 (2017-12-20)
S C Gross et al.
Orthopedics, 12(12), 1561-1564 (1989-12-01)
From 1968 to 1987, 22 patients were diagnosed with dysfunctioning digits after complete distal digital amputations. Each patient had the proximal portion of the partially amputated phalanx left within the injured digit. On average, 21 months after the initial injury

Questions

  1. This is 100% Anamorelin? 5mg each capsule? Thanks 

    1 answer
    1. This product has a minimum purity specification of 97.5%. There may be some moisture present which is reported on the lot specific Certificate of Analysis. Note that material is not in capsule form. It is a loose powder. Unless otherwise stated this product is intended for research use only and is not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals. Please see the link below to review a sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/SML2519-25MG0000192446.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service