Skip to Content
Merck
All Photos(1)

Documents

SML3680

Sigma-Aldrich

Macimorelin acetate

≥98% (HPLC)

Synonym(s):

2-Amino-N-[(2R)-1-[[(1R)-1-formamido-2-(1H-indol-3-yl)ethyl]amino]-3-1H-indol-3-yl)-1-oxopropan-2-yl]-2-methylpropanamide acetate, 2-Methylalanyl-N-[(1R)-1-(formylamino)-2-(1H-indol-3-yl)ethyl]-D-tryptophanamide acetate, AEZS 130 acetate, AEZS-130 acetate, AEZS130 acetate, ARD 07 acetate, ARD-07 acetate, ARD07 acetate, Aib-DTrp-DgTrp-CHO acetate, EP 1572 acetate, EP-1572 acetate, EP1572 acetate, JMV 1843 acetate, JMV-1843 acetate, JMV1843 acetate, UMV 1843 acetate, UMV-1843 acetate, UMV1843 acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30N6O3 · xC2H4O2
Molecular Weight:
474.55 (free base basis)
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Macimorelin (AEZS-130; ARD-07; EP1572; UMV1843; Aib-DTrp-DgTrp-CHO) is an orally active peptido-mimetic growth hormone (GH) secretagogue (GHS) that targets human and animal GHS-receptor (IC50 in nM = 10.2/ghrelin, 12.3/hexaghrelin, 15.6/macimorelin against ghrelin binding to human GHS-R) with potent and selective GH-releasing activity in vivo (GH level = 158.8 μg/L 15 min post 300 μg/kg s.c. in neonatal rats; 11.3 μg/L without treatment).

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ghrelin stimulation of growth hormone-releasing hormone neurons is direct in the arcuate nucleus
PLoS ONE, 5(2), e9159-e9159 (2010)
F Broglio et al.
Journal of endocrinological investigation, 25(8), RC26-RC28 (2002-09-21)
EP1572 UMV1843 [Aib-DTrp-DgTrp-CHO]) is a new peptido-mimetic GH secretagogue (GHS) showing binding potency to the GHS-receptor in animal and human tissues similar to that of ghrelin and peptidyl GHS. EP1572 induces marked GH increase after s.c. administration in neonatal rats.
Valeria Sibilia et al.
Amino acids, 43(4), 1751-1759 (2012-03-13)
Recent research suggests a role for ghrelin in the modulation of inflammatory disorders. However, the type of ghrelin receptor (GHS-R) involved in both the anti-inflammatory and anti-hyperalgesic actions of ghrelin remains to be characterized. In this study, we examined whether
M Holubová et al.
Physiological research, 62(4), 435-444 (2013-04-18)
Ghrelin and agonists of its receptor GHS-R1a are potential substances for the treatment of cachexia. In the present study, we investigated the acute and long term effects of the GHS R1a agonist JMV 1843 (H Aib-DTrp-D-gTrp-CHO) on food intake, body
An Buckinx et al.
European journal of neurology, 28(9), 3100-3112 (2021-06-23)
Current drugs for epilepsy affect seizures, but no antiepileptogenic or disease-modifying drugs are available that prevent or slow down epileptogenesis, which is characterized by neuronal cell loss, inflammation and aberrant network formation. Ghrelin and ghrelin receptor (ghrelin-R) agonists were previously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service