Skip to Content
Merck
All Photos(1)

Key Documents

SML2510

Sigma-Aldrich

Flubendazol

Synonym(s):

Flubendazol, Flubendazole, Flumoxanal, Methyl [5-(4-fluorobenzoyl)-1H-benzimidazol-2-yl]carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$367.00

$367.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$367.00

About This Item

Empirical Formula (Hill Notation):
C16H12FN3O3
CAS Number:
Molecular Weight:
313.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$367.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

storage temp.

2-8°C

SMILES string

COC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccc(F)cc3

InChI

1S/C16H12FN3O3/c1-23-16(22)20-15-18-12-7-4-10(8-13(12)19-15)14(21)9-2-5-11(17)6-3-9/h2-8H,1H3,(H2,18,19,20,22)

InChI key

CPEUVMUXAHMANV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Flubendazole, an antithelmintic, has been widely used in treating intestinal parasites. Additionally, Flubendazole has been reported to exert anticancer activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongqi Zhen et al.
Theranostics, 10(18), 8080-8097 (2020-07-30)
Background: Triple-negative breast cancer (TNBC) is one of the most prevalent neoplastic diseases worldwide, but efficacious treatments for this pathological condition are still challenging. The lack of an effective targeted therapy also leads to a poor prognosis for patients affected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service