Skip to Content
Merck

Skip To

SML2425

Manidipine hydrochloride

≥98% (HPLC)

Synonym(s):

1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid hydrchloride, 2-[4-(diphenylmethyl)-1-piperazinyl]ethyl methyl ester, 3-(2-(4-Benzhydrylpiperazin-1-yl)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrchloride, CV 4093 hydrchloride, CV-4093 hydrchloride, CV4093 hydrchloride, Franidipine hydrchloride, Manidipine 6300 hydrchloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$115.00
25 mg
Check Cart for Availability
$469.00

About This Item

Empirical Formula (Hill Notation):
C35H38N4O6 · xHCl
CAS Number:
Molecular Weight:
610.70 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated

$115.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Dihydropyridine class calcium channel blocker (CCB) that exerits its antihypertensive activity via targeting both L-type and T-type calcium channels.
Manidipine is a dihydropyridine class calcium channel blocker (CCB) that exerits its antihypertensive activity via targeting both L-type and T-type calcium channels. In contrary to other dihydropyridines and non-dihydropyridines that mainly act as L-type CCBs, Manidipine diminishes glomerular pressure and, consequently, albumin excretion via its action against T-type channels of efferent arterioles in addition to promoting afferent arteriole dilation by blocking L-type channels. Clinically, Manidipine is as effective in lowering blood pressure as other dihydropyridines, while only Manidipine significantly reduces albuminuria and insulin resistance with less adverse effects. In obese and hypertensive individuals, Manidipine is more effective than Lercanidipine in reducing insulin resistance, while Nifedipine treatment is reported to increase insulin desensitisation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B6813N7510L6668
description

≥98% (HPLC)

description

≥98% (HPLC)

description

powder, ≥98%

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98%

assay

≥98% (HPLC)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear (warmed)

solubility

DMSO: ≥20 mg/mL

solubility

DMSO: ~1 mg/mL, 0.1 M NaOH: insoluble, H2O: slightly soluble, methanol: soluble

solubility

-

storage condition

desiccated

storage condition

desiccated

storage condition

-

storage condition

desiccated, protect from light

color

white to beige

color

yellow

color

yellow

color

yellow


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters