Skip to Content
Merck
All Photos(1)

Documents

B6813

Sigma-Aldrich

Benidipine hydrochloride

≥98% (HPLC)

Synonym(s):

(4R)-rel-3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(3-nitrophenyl)-, 3-methyl 5-[(3R)-1-(phenylmethyl)-3-piperidinyl] ester, KW-3049, hydrochloride (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31N3O6·HCl
CAS Number:
Molecular Weight:
542.02
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

yellow

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

Cl.COC(=O)C1=C(C)NC(C)=C([C@@H]1c2cccc(c2)[N+]([O-])=O)C(=O)O[C@@H]3CCCN(C3)Cc4ccccc4

InChI

1S/C28H31N3O6.ClH/c1-18-24(27(32)36-3)26(21-11-7-12-22(15-21)31(34)35)25(19(2)29-18)28(33)37-23-13-8-14-30(17-23)16-20-9-5-4-6-10-20;/h4-7,9-12,15,23,26,29H,8,13-14,16-17H2,1-3H3;1H/t23-,26-;/m1./s1

InChI key

KILKDKRQBYMKQX-MIPPOABVSA-N

Biochem/physiol Actions

Calcium Channel Blocker

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María L Sbaraglini et al.
International journal of antimicrobial agents, 48(1), 91-95 (2016-05-25)
Despite current efforts worldwide to develop new medications against Chagas disease, only two drugs are available, nifurtimox and benznidazole. Both drugs require prolonged treatment and have multiple side effects and limited efficacy on adult patients chronically infected with Trypanosoma cruzi.
Takatsugu Oike et al.
Scientific reports, 7(1), 10965-10965 (2017-09-10)
Rheumatoid arthritis (RA) is a multi-factorial disease characterized by chronic inflammation and destruction of multiple joints. To date, various biologic treatments for RA such as anti-tumor necrosis factor alpha antibodies have been developed; however, mechanisms underlying RA development remain unclear
Noriaki Ikemura et al.
Chemico-biological interactions, 306, 1-9 (2019-04-10)
The inhibitory effects of antihypertensive drugs (dihydropyridine calcium channel blockers, angiotensin II receptor blockers, and angiotensin-converting enzyme inhibitors) on cytochrome P450 2J2 (CYP2J2) activity were examined. Amlodipine, azelnidipine, barnidipine, benidipine, cilnidipine, efonidipine, felodipine, manidipine, nicardipine, nifedipine, nilvadipine, nisoldipine, nitrendipine, telmisartan

Articles

We offers many products related to calcium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service