Skip to Content
Merck
All Photos(1)

Key Documents

SML2379

Sigma-Aldrich

Fluorizoline

≥98% (HPLC)

Synonym(s):

2,5-bis(4-Chlorophenyl)-4,4,5-trifluoro-4,5-dihydrothiazole, 4,4,5-Trifluoro-2,5-bis(4-chlorophenyl)-4,5-dihydrothiazole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$118.00
25 MG
$473.00

$118.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$118.00
25 MG
$473.00

About This Item

Empirical Formula (Hill Notation):
C15H8Cl2F3NS
CAS Number:
Molecular Weight:
362.20
UNSPSC Code:
12352200
NACRES:
NA.77

$118.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC=C1C2=NC(F)(F)C(C3=CC=C(Cl)C=C3)(F)S2

InChI key

JGDLZQYCDVDWNE-UHFFFAOYSA-N

Biochem/physiol Actions

Fluorizoline is a prohibitin (PHB) 1/2-binding diaryl trifluorothiazoline compound that induces p53-independent apoptosis in various cancer cultures (IC50 from 1.6 μM/Bx-PC3 to 9.4 μM/JeKo 1; 24 h) and upregulates proapoptotic Noxa and Bim mRNAs in wild-type, but not PHBs-depleted MEFs. Box(-/-)/Bak(-/-) or Noxa(-/-)/Bim(-/-) MEFs as well as BAX/BAK- or NOXA-downregulated HeLa cells are resistant to fluorizoline-induced apoptosis. Fluorizoline treatment is shown to prevent active RAS-CRAF interaction and downstream MEK1/2 activation following EGF stimulation in HeLa cells (10 μM).
Prohibitin (PHB) 1/2-binding diaryl trifluorothiazoline that induces p53-independent apoptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alba Pérez-Perarnau et al.
Angewandte Chemie (International ed. in English), 53(38), 10150-10154 (2014-09-10)
A new class of small molecules, with an unprecedented trifluorothiazoline scaffold, were synthesized and their pro-apoptotic activity was evaluated. With an EC50 in the low micromolar range, these compounds proved to be potent inducers of apoptosis in a broad spectrum
Ana M Cosialls et al.
Haematologica, 102(9), 1587-1593 (2017-06-18)
Fluorizoline is a new synthetic molecule that induces apoptosis by selectively targeting prohibitins. In the study herein, the pro-apoptotic effect of fluorizoline was assessed in 34 primary samples from patients with chronic lymphocytic leukemia. Fluorizoline induced apoptosis in chronic lymphocytic
The prohibitin-binding compound fluorizoline induces apoptosis in chronic lymphocytic leukemia cells ex vivo but fails to prevent leukemia development in a murine model.
Marina Wierz et al.
Haematologica, 103(4), e154-e157 (2018-01-07)
H Yurugi et al.
Oncogene, 36(33), 4778-4789 (2017-04-18)
KRAS is one of the most frequently mutated oncogenes in human non-small cell lung cancers (NSCLCs). RAS proteins trigger multiple effector signalling pathways including the highly conserved RAF-MAPK pathway. CRAF, a direct RAS effector protein, is required for KRAS-mediated tumourigenesis.
Helena Pomares et al.
Oncotarget, 7(40), 64987-65000 (2016-08-20)
Fluorizoline is a new synthetic molecule that induces apoptosis by selectively targeting prohibitins (PHBs). In this study, the pro-apoptotic effect of fluorizoline was assessed in two cell lines and 21 primary samples from patients with debut of acute myeloid leukemia

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service