Skip to Content
Merck
All Photos(1)

Key Documents

SML2289

Sigma-Aldrich

HBT1

Synonym(s):

2-(((5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl)acetyl)amino)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide, N-[3-(Aminocarbonyl)-4,5,6,7-tetrahydrobenzo[b]thien-2-yl]-5-methyl-3-(trifluoromethyl)-1H-pyrazole-1-acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$109.00
25 MG
$439.00

$109.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$109.00
25 MG
$439.00

About This Item

Empirical Formula (Hill Notation):
C16H17F3N4O2S
CAS Number:
Molecular Weight:
386.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$109.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N)C1=C(NC(CN2C(C)=CC(C(F)(F)F)=N2)=O)SC3=C1CCCC3

InChI

1S/C16H17F3N4O2S/c1-8-6-11(16(17,18)19)22-23(8)7-12(24)21-15-13(14(20)25)9-4-2-3-5-10(9)26-15/h6H,2-5,7H2,1H3,(H2,20,25)(H,21,24)

InChI key

PHLXSNIEQIKENK-UHFFFAOYSA-N

Biochem/physiol Actions

AMPA-R potentiator that induces production of brain-derived neurotrophic factor (BDNF) and exhibits little agonistic effect in primary neurons
HBT1 is an AMPA-R [alpha-amino-3-hydroxy-5-methyl-4-isoxazole-propionic acid (AMPA) receptor] potentiator that induces production of brain-derived neurotrophic factor (BDNF) and exhibits little agonistic effect in primary neurons. HBT1 binds to ligand-binding domain of AMPA-R in glutamate dependent manner.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akiyoshi Kunugi et al.
The Journal of pharmacology and experimental therapeutics, 364(3), 377-389 (2018-01-05)
α-Amino-3-hydroxy-5-methyl-4-isoxazole-propionic acid (AMPA) receptor (AMPA-R) potentiators with brain-derived neurotrophic factor (BDNF)-induction potential could be promising as therapeutic drugs for neuropsychiatric and neurologic disorders. However, AMPA-R potentiators such as LY451646 have risks of narrow bell-shaped responses in pharmacological effects, including in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service